Assay Details
| Gene Symbol: MGAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002409.4 | NP_002400.3 | 1 - 2 | 238 | 140 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000315.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_002409.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR22005937.hg.1 | ENST00000341184.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22005937.hg.1 | NM_002409.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22005937.hg.1 | TC22000315.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22005943.hg.1 | ENST00000341184.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22005943.hg.1 | NM_001098270.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22005943.hg.1 | NM_002409.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22005943.hg.1 | TC22000315.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22005945.hg.1 | ENST00000341184.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22005945.hg.1 | NM_001098270.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22005945.hg.1 | NM_002409.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22005945.hg.1 | TC22000315.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007402.hg.1 | ENST00000341184.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007402.hg.1 | ENST00000341184.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007402.hg.1 | ENST00000341184.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007402.hg.1 | ENST00000341184.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007402.hg.1 | ENST00000341184.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007402.hg.1 | ENST00000341184.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007402.hg.1 | ENST00000341184.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007402.hg.1 | ENST00000341184.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007402.hg.1 | ENST00000418314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007402.hg.1 | ENST00000418314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007402.hg.1 | ENST00000418314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007402.hg.1 | ENST00000429402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007402.hg.1 | ENST00000429402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007402.hg.1 | ENST00000429402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007402.hg.1 | NM_001098270.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007402.hg.1 | NM_001098270.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007402.hg.1 | NM_001098270.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007402.hg.1 | NM_001098270.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007402.hg.1 | NM_001098270.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007402.hg.1 | NM_001098270.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007402.hg.1 | NM_001098270.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007402.hg.1 | NM_002409.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007402.hg.1 | NM_002409.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007402.hg.1 | NM_002409.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007402.hg.1 | NM_002409.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007402.hg.1 | NM_002409.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007402.hg.1 | NM_002409.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007402.hg.1 | NM_002409.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007402.hg.1 | NM_002409.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22005944.hg.1 | ENST00000341184.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22005944.hg.1 | NM_001098270.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22005944.hg.1 | NM_002409.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22005944.hg.1 | TC22000315.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC2200007402.hg.1 | ENST00000341184.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007402.hg.1 | ENST00000341184.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007402.hg.1 | ENST00000341184.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007402.hg.1 | ENST00000341184.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007402.hg.1 | ENST00000341184.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007402.hg.1 | ENST00000341184.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007402.hg.1 | ENST00000341184.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007402.hg.1 | ENST00000341184.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007402.hg.1 | ENST00000418314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007402.hg.1 | ENST00000418314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007402.hg.1 | ENST00000418314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007402.hg.1 | ENST00000429402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007402.hg.1 | ENST00000429402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007402.hg.1 | ENST00000429402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007402.hg.1 | NM_001098270.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007402.hg.1 | NM_001098270.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007402.hg.1 | NM_001098270.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007402.hg.1 | NM_001098270.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007402.hg.1 | NM_001098270.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007402.hg.1 | NM_001098270.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007402.hg.1 | NM_001098270.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007402.hg.1 | NM_002409.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007402.hg.1 | NM_002409.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007402.hg.1 | NM_002409.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007402.hg.1 | NM_002409.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007402.hg.1 | NM_002409.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007402.hg.1 | NM_002409.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007402.hg.1 | NM_002409.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007402.hg.1 | NM_002409.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200152174.hg.1 | TC2200007402.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200152174.hg.1 | ENST00000341184.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200152174.hg.1 | NM_002409.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200152174.hg.1 | ENST00000341184.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200152174.hg.1 | NM_002409.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200152181.hg.1 | TC2200007402.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200152181.hg.1 | ENST00000341184.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200152181.hg.1 | ENST00000418314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200152181.hg.1 | ENST00000429402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200152181.hg.1 | NM_001098270.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200152181.hg.1 | NM_002409.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200152181.hg.1 | ENST00000341184.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200152181.hg.1 | ENST00000418314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200152181.hg.1 | ENST00000429402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200152181.hg.1 | NM_001098270.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200152181.hg.1 | NM_002409.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200152182.hg.1 | TC2200007402.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200152182.hg.1 | ENST00000341184.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200152182.hg.1 | ENST00000429402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200152182.hg.1 | NM_001098270.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200152182.hg.1 | NM_002409.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200152182.hg.1 | ENST00000341184.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200152182.hg.1 | ENST00000429402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200152182.hg.1 | NM_001098270.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200152182.hg.1 | NM_002409.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200152175.hg.1 | TC2200007402.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200152175.hg.1 | ENST00000341184.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200152175.hg.1 | NM_002409.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200152175.hg.1 | ENST00000341184.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200152175.hg.1 | NM_002409.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200152183.hg.1 | TC2200007402.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200152183.hg.1 | ENST00000341184.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200152183.hg.1 | NM_001098270.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200152183.hg.1 | NM_002409.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200152183.hg.1 | ENST00000341184.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200152183.hg.1 | NM_001098270.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200152183.hg.1 | NM_002409.4 | Refseq |

MGAT3
Genomic Map