Assay Details
| Gene Symbol: FMO6P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002601.1 | 2 - 3 | 944 | 82 | ||
| GenBank mRNA | AK130511.1 | - | 2 - 3 | 944 | 82 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | flavin containing monooxygenase 3 | FMO3 |
FMOII, TMAU, dJ127D3.1 |
-28.7kb | Sense | Chr. 1 - 171090873 - 171117819 on NCBI Build GRCh38 |
| Downstream | flavin containing monooxygenase 2 | FMO2 | FMO1B1 | 37.4kb | Sense | Chr. 1 - 171185208 - 171212683 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001487.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01023060.hg.1 | TC01001487.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01023060.hg.1 | NR_002601.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01023066.hg.1 | TC01001487.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01023066.hg.1 | ENST00000236166.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01023066.hg.1 | NR_002601.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01023062.hg.1 | TC01001487.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01023062.hg.1 | NR_002601.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01023061.hg.1 | TC01001487.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01023061.hg.1 | ENST00000236166.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01023061.hg.1 | NR_002601.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010581.hg.1 | ENST00000236166.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010581.hg.1 | ENST00000236166.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010581.hg.1 | ENST00000236166.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010581.hg.1 | ENST00000236166.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010581.hg.1 | ENST00000236166.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010581.hg.1 | ENST00000236166.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010581.hg.1 | ENST00000236166.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010581.hg.1 | ENST00000236166.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010581.hg.1 | ENST00000236166.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010581.hg.1 | ENST00000236166.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010581.hg.1 | ENST00000236166.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010581.hg.1 | ENST00000367754.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010581.hg.1 | ENST00000367754.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010581.hg.1 | ENST00000367754.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010581.hg.1 | ENST00000367754.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010581.hg.1 | ENST00000367754.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010581.hg.1 | ENST00000367754.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010581.hg.1 | ENST00000367754.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010581.hg.1 | ENST00000367754.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010581.hg.1 | ENST00000367754.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010581.hg.1 | ENST00000367754.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010581.hg.1 | ENST00000367754.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010581.hg.1 | ENST00000633725.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010581.hg.1 | ENST00000633725.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010581.hg.1 | ENST00000633725.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010581.hg.1 | NR_002601.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010581.hg.1 | NR_002601.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010581.hg.1 | NR_002601.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010581.hg.1 | NR_002601.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010581.hg.1 | NR_002601.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010581.hg.1 | NR_002601.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010581.hg.1 | NR_002601.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010581.hg.1 | NR_002601.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010581.hg.1 | NR_002601.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010581.hg.1 | NR_002601.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010581.hg.1 | NR_002601.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01023065.hg.1 | TC01001487.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01023065.hg.1 | ENST00000236166.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01023065.hg.1 | NR_002601.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100177877.hg.1 | ENST00000367754.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177877.hg.1 | NR_002601.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100177877.hg.1 | ENST00000367754.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177877.hg.1 | NR_002601.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100177877.hg.1 | TC0100010581.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100177878.hg.1 | ENST00000236166.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177878.hg.1 | ENST00000367754.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177878.hg.1 | NR_002601.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100177878.hg.1 | ENST00000236166.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177878.hg.1 | ENST00000367754.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177878.hg.1 | NR_002601.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100177878.hg.1 | TC0100010581.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100177879.hg.1 | ENST00000367754.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177879.hg.1 | NR_002601.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100177879.hg.1 | ENST00000367754.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177879.hg.1 | NR_002601.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100177879.hg.1 | TC0100010581.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100177882.hg.1 | ENST00000236166.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177882.hg.1 | ENST00000367754.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177882.hg.1 | NR_002601.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100177882.hg.1 | ENST00000236166.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177882.hg.1 | ENST00000367754.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177882.hg.1 | NR_002601.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100177882.hg.1 | TC0100010581.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100177883.hg.1 | ENST00000236166.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177883.hg.1 | ENST00000367754.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177883.hg.1 | NR_002601.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100177883.hg.1 | ENST00000236166.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177883.hg.1 | ENST00000367754.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177883.hg.1 | NR_002601.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100177883.hg.1 | TC0100010581.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100010581.hg.1 | ENST00000236166.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010581.hg.1 | ENST00000236166.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010581.hg.1 | ENST00000236166.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010581.hg.1 | ENST00000236166.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010581.hg.1 | ENST00000236166.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010581.hg.1 | ENST00000236166.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010581.hg.1 | ENST00000236166.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010581.hg.1 | ENST00000236166.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010581.hg.1 | ENST00000236166.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010581.hg.1 | ENST00000236166.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010581.hg.1 | ENST00000236166.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010581.hg.1 | ENST00000367754.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010581.hg.1 | ENST00000367754.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010581.hg.1 | ENST00000367754.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010581.hg.1 | ENST00000367754.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010581.hg.1 | ENST00000367754.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010581.hg.1 | ENST00000367754.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010581.hg.1 | ENST00000367754.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010581.hg.1 | ENST00000367754.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010581.hg.1 | ENST00000367754.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010581.hg.1 | ENST00000367754.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010581.hg.1 | ENST00000367754.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010581.hg.1 | ENST00000633725.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010581.hg.1 | ENST00000633725.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010581.hg.1 | ENST00000633725.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010581.hg.1 | NR_002601.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010581.hg.1 | NR_002601.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010581.hg.1 | NR_002601.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010581.hg.1 | NR_002601.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010581.hg.1 | NR_002601.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010581.hg.1 | NR_002601.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010581.hg.1 | NR_002601.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010581.hg.1 | NR_002601.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010581.hg.1 | NR_002601.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010581.hg.1 | NR_002601.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010581.hg.1 | NR_002601.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100177876.hg.1 | ENST00000367754.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177876.hg.1 | NR_002601.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100177876.hg.1 | ENST00000367754.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177876.hg.1 | NR_002601.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100177876.hg.1 | TC0100010581.hg.1 | Clariom D TC |

FMO6P
Genomic Map