| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010581.hg.1 |
ENST00000236166.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010581.hg.1 |
ENST00000236166.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010581.hg.1 |
ENST00000236166.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010581.hg.1 |
ENST00000236166.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010581.hg.1 |
ENST00000236166.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010581.hg.1 |
ENST00000236166.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010581.hg.1 |
ENST00000236166.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010581.hg.1 |
ENST00000236166.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010581.hg.1 |
ENST00000236166.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010581.hg.1 |
ENST00000236166.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010581.hg.1 |
ENST00000236166.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010581.hg.1 |
ENST00000367754.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010581.hg.1 |
ENST00000367754.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010581.hg.1 |
ENST00000367754.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010581.hg.1 |
ENST00000367754.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010581.hg.1 |
ENST00000367754.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010581.hg.1 |
ENST00000367754.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010581.hg.1 |
ENST00000367754.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010581.hg.1 |
ENST00000367754.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010581.hg.1 |
ENST00000367754.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010581.hg.1 |
ENST00000367754.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010581.hg.1 |
ENST00000367754.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010581.hg.1 |
ENST00000633725.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010581.hg.1 |
ENST00000633725.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010581.hg.1 |
ENST00000633725.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010581.hg.1 |
NR_002601.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010581.hg.1 |
NR_002601.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010581.hg.1 |
NR_002601.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010581.hg.1 |
NR_002601.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010581.hg.1 |
NR_002601.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010581.hg.1 |
NR_002601.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010581.hg.1 |
NR_002601.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010581.hg.1 |
NR_002601.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010581.hg.1 |
NR_002601.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010581.hg.1 |
NR_002601.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010581.hg.1 |
NR_002601.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01023069.hg.1 |
TC01001487.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01023069.hg.1 |
ENST00000236166.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01023069.hg.1 |
NR_002601.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100177886.hg.1 |
ENST00000236166.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100177886.hg.1 |
ENST00000367754.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100177886.hg.1 |
NR_002601.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100177886.hg.1 |
ENST00000236166.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100177886.hg.1 |
ENST00000367754.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100177886.hg.1 |
NR_002601.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100177886.hg.1 |
TC0100010581.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100010581.hg.1 |
ENST00000236166.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010581.hg.1 |
ENST00000236166.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010581.hg.1 |
ENST00000236166.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010581.hg.1 |
ENST00000236166.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010581.hg.1 |
ENST00000236166.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010581.hg.1 |
ENST00000236166.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010581.hg.1 |
ENST00000236166.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010581.hg.1 |
ENST00000236166.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010581.hg.1 |
ENST00000236166.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010581.hg.1 |
ENST00000236166.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010581.hg.1 |
ENST00000236166.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010581.hg.1 |
ENST00000367754.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010581.hg.1 |
ENST00000367754.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010581.hg.1 |
ENST00000367754.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010581.hg.1 |
ENST00000367754.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010581.hg.1 |
ENST00000367754.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010581.hg.1 |
ENST00000367754.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010581.hg.1 |
ENST00000367754.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010581.hg.1 |
ENST00000367754.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010581.hg.1 |
ENST00000367754.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010581.hg.1 |
ENST00000367754.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010581.hg.1 |
ENST00000367754.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010581.hg.1 |
ENST00000633725.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010581.hg.1 |
ENST00000633725.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010581.hg.1 |
ENST00000633725.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010581.hg.1 |
NR_002601.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010581.hg.1 |
NR_002601.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010581.hg.1 |
NR_002601.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010581.hg.1 |
NR_002601.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010581.hg.1 |
NR_002601.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010581.hg.1 |
NR_002601.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010581.hg.1 |
NR_002601.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010581.hg.1 |
NR_002601.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010581.hg.1 |
NR_002601.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010581.hg.1 |
NR_002601.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010581.hg.1 |
NR_002601.1 |
Refseq |