Assay Details
| Gene Symbol: LINC00558 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_047488.1 | 5 - 6 | 489 | 149 | ||
| GenBank mRNA | BC041877.1 | - | 5 - 6 | 489 | 149 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1065 | LINC01065 | -718.6kb | Anti-sense | Chr. 13 - 53131746 - 53151900 on NCBI Build GRCh38 | |
| Downstream | long intergenic non-protein coding RNA 458 | LINC00458 | LncRNA-ES3 | 248.2kb | Anti-sense | Chr. 13 - 54124324 - 54132871 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LINC00558
Genomic Map