Assay Details
| Gene Symbol: BMS1P20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027293.2 | 9 - 10 | 1416 | 71 | ||
| GenBank mRNA | AK090629.1 | - | 9 - 10 | 1356 | 71 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | pre-B lymphocyte 1 | VPREB1 |
CD179a, IGI, IGVPB, VPREB |
-73.4kb | Sense | Chr. 22 - 22244780 - 22245515 on NCBI Build GRCh38 |
| Downstream | immunoglobulin lambda variable 1-44 | IGLV1-44 |
IGLV144, V1-16 |
59.0kb | Sense | Chr. 22 - 22380880 - 22381347 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22001450.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000390289.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000390289.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000390290.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000390290.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000390290.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000413293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000413293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000413293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000413293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000413293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000413293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000413293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000413293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000413293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000413293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000413293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000436538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000436538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000436538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22001550.hg.1 | TC22001450.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22001550.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22001550.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000390289.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000390289.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000390290.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000390290.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000390290.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000413293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000413293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000413293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000413293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000413293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000413293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000413293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000413293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000413293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000413293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000413293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000436538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000436538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000436538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200167635.hg.1 | TC2200009206.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200167635.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200167635.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200167635.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200167635.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200167641.hg.1 | TC2200009206.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200167641.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200167641.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200167641.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200167641.hg.1 | NR_027293.2 | Refseq |

BMS1P20
Genomic Map