Assay Details
| Gene Symbol: GAB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037814.1 | NP_001032903.1 | 3 - 4 | 793 | 57 | |
| XM_011546116.2 | XP_011544418.1 | 3 - 4 | 852 | 57 | ||
| XM_017028575.1 | XP_016884064.1 | 3 - 4 | 845 | 57 | ||
| GenBank mRNA | AB076978.1 | - | 3 - 4 | 793 | 57 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000469.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR22008401.hg.1 | ENST00000400588.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008401.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008401.hg.1 | TC22000469.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008405.hg.1 | ENST00000400588.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008405.hg.1 | ENST00000523144.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008405.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008405.hg.1 | TC22000469.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008404.hg.1 | ENST00000400588.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008404.hg.1 | ENST00000523144.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008404.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008404.hg.1 | TC22000469.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008402.hg.1 | ENST00000400588.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008402.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008402.hg.1 | TC22000469.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008403.hg.1 | ENST00000400588.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008403.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008403.hg.1 | TC22000469.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008406.hg.1 | ENST00000400588.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008406.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008406.hg.1 | TC22000469.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | ENST00000465611.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | ENST00000465611.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | ENST00000465611.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | ENST00000465611.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | ENST00000465611.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | ENST00000465611.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | ENST00000465611.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | ENST00000465611.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | ENST00000465611.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | ENST00000465611.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | ENST00000465611.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | ENST00000465611.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | ENST00000465611.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | ENST00000465611.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | ENST00000520505.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | ENST00000520505.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | ENST00000523144.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | ENST00000523144.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | ENST00000523144.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | ENST00000523144.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009310.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008396.hg.1 | ENST00000400588.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008396.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008396.hg.1 | TC22000469.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008400.hg.1 | ENST00000400588.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008400.hg.1 | ENST00000523144.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008400.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008400.hg.1 | TC22000469.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2200169652.hg.1 | TC2200009310.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200169652.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169652.hg.1 | ENST00000465611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169652.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200169652.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169652.hg.1 | ENST00000465611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169652.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200169660.hg.1 | TC2200009310.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200169660.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169660.hg.1 | ENST00000465611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169660.hg.1 | ENST00000523144.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169660.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200169660.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169660.hg.1 | ENST00000465611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169660.hg.1 | ENST00000523144.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169660.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | ENST00000465611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | ENST00000465611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | ENST00000465611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | ENST00000465611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | ENST00000465611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | ENST00000465611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | ENST00000465611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | ENST00000465611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | ENST00000465611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | ENST00000465611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | ENST00000465611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | ENST00000465611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | ENST00000465611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | ENST00000465611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | ENST00000520505.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | ENST00000520505.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | ENST00000523144.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | ENST00000523144.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | ENST00000523144.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | ENST00000523144.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009310.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200169657.hg.1 | TC2200009310.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200169657.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169657.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200169657.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169657.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200169661.hg.1 | TC2200009310.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200169661.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169661.hg.1 | ENST00000465611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169661.hg.1 | ENST00000523144.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169661.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200169661.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169661.hg.1 | ENST00000465611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169661.hg.1 | ENST00000523144.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169661.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200169656.hg.1 | TC2200009310.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200169656.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169656.hg.1 | ENST00000465611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169656.hg.1 | ENST00000520505.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169656.hg.1 | ENST00000523144.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169656.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200169656.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169656.hg.1 | ENST00000465611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169656.hg.1 | ENST00000520505.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169656.hg.1 | ENST00000523144.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169656.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200169663.hg.1 | TC2200009310.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200169663.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169663.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200169663.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169663.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200169658.hg.1 | TC2200009310.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200169658.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169658.hg.1 | ENST00000465611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169658.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200169658.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169658.hg.1 | ENST00000465611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169658.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200169659.hg.1 | TC2200009310.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200169659.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169659.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200169659.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169659.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200169662.hg.1 | TC2200009310.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200169662.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169662.hg.1 | NM_001037814.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200169662.hg.1 | ENST00000400588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169662.hg.1 | NM_001037814.1 | Refseq |

GAB4
Genomic Map