Assay Details
| Gene Symbol: MIRLET7DHG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046163.1 | 3 - 4 | 185 | 76 | ||
| NR_046165.1 | 2 - 3 | 126 | 76 | |||
| GenBank mRNA | BC036695.1 | - | 3 - 4 | 164 | 76 | |
| BC045813.1 | - | 2 - 3 | 126 | 76 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001354.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06825783 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06825777 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06825788 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27167246 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16242363 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06825785 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06825794 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27167244 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27167242 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06825772 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06825789 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06825774 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06825784 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06825795 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27167231 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06825778 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06825793 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06825786 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06825787 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06825773 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06825792 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27167240 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27167261 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27167256 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27167260 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27167248 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 2085 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 56964 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 177607 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 180652 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 8x60K | 38483 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 420635 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 384352 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 119903 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 843036 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 561222 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 860146 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 624626 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 124918 | BC112212.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 124918 | BC113633.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 265322 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 763038 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 79506 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 12971 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 230126 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 152075 | U39412.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 109918 | AY183470.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 109918 | BC069157.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 109918 | NM_175067.1 | refseq_mrna_human_NM |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3417740 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 21411 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3417740 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P18698002 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P02134909 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P02134909 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P18697985 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P18698002 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P38798335 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 137338 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 38736 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 15259 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 48183 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02134904 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P38798335 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P18697996 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P18697985 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P18697977 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P16924967 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P18698002 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02134903 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P18698005 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P26649130 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P16921498 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P18698000 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P38798318 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02134909 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P382881 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P18698002 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P02134909 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_18_P16921498 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_18_P16924967 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P38798335 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P18697985 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 1559129_PM_a_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR09016913.hg.1 | TC09001354.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09016913.hg.1 | ENST00000416309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09016913.hg.1 | NR_046163.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09016913.hg.1 | NR_046165.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010850.hg.1 | ENST00000416309.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010850.hg.1 | ENST00000416309.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010850.hg.1 | ENST00000416309.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010850.hg.1 | ENST00000416309.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010850.hg.1 | ENST00000416309.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010850.hg.1 | ENST00000416309.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010850.hg.1 | ENST00000416309.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010850.hg.1 | ENST00000416309.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010850.hg.1 | ENST00000602602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010850.hg.1 | ENST00000602602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010850.hg.1 | ENST00000602602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010850.hg.1 | NR_046163.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010850.hg.1 | NR_046163.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010850.hg.1 | NR_046163.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010850.hg.1 | NR_046163.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010850.hg.1 | NR_046163.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010850.hg.1 | NR_046165.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010850.hg.1 | NR_046165.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010850.hg.1 | NR_046165.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010850.hg.1 | NR_046165.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09016911.hg.1 | TC09001354.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09016911.hg.1 | ENST00000416309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09016911.hg.1 | NR_046163.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09016911.hg.1 | NR_046165.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09016914.hg.1 | TC09001354.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09016914.hg.1 | NR_046163.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09016914.hg.1 | NR_046165.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09016912.hg.1 | TC09001354.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09016912.hg.1 | ENST00000416309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09016912.hg.1 | NR_046163.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09016908.hg.1 | TC09001354.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09016908.hg.1 | NR_046163.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09016908.hg.1 | NR_046165.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09016910.hg.1 | TC09001354.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09016910.hg.1 | ENST00000416309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09016910.hg.1 | NR_046163.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09016910.hg.1 | NR_046165.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900173793.hg.1 | ENST00000416309.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900173793.hg.1 | ENST00000416309.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900173793.hg.1 | ENST00000602602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900173793.hg.1 | NR_046163.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900173793.hg.1 | NR_046165.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900173793.hg.1 | ENST00000416309.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900173793.hg.1 | ENST00000416309.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900173793.hg.1 | ENST00000602602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900173793.hg.1 | NR_046163.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900173793.hg.1 | NR_046165.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900173793.hg.1 | TC0900010850.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900010850.hg.1 | ENST00000416309.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010850.hg.1 | ENST00000416309.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010850.hg.1 | ENST00000416309.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010850.hg.1 | ENST00000416309.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010850.hg.1 | ENST00000416309.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010850.hg.1 | ENST00000416309.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010850.hg.1 | ENST00000416309.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010850.hg.1 | ENST00000416309.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010850.hg.1 | ENST00000602602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010850.hg.1 | ENST00000602602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010850.hg.1 | ENST00000602602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010850.hg.1 | NR_046163.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010850.hg.1 | NR_046163.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010850.hg.1 | NR_046163.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010850.hg.1 | NR_046163.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010850.hg.1 | NR_046163.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010850.hg.1 | NR_046165.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010850.hg.1 | NR_046165.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010850.hg.1 | NR_046165.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010850.hg.1 | NR_046165.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900173792.hg.1 | NR_046163.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900173792.hg.1 | NR_046165.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900173792.hg.1 | NR_046163.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900173792.hg.1 | NR_046165.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900173792.hg.1 | TC0900010850.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900173796.hg.1 | ENST00000416309.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900173796.hg.1 | ENST00000416309.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900173796.hg.1 | ENST00000602602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900173796.hg.1 | NR_046163.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900173796.hg.1 | NR_046165.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900173796.hg.1 | ENST00000416309.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900173796.hg.1 | ENST00000416309.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900173796.hg.1 | ENST00000602602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900173796.hg.1 | NR_046163.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900173796.hg.1 | NR_046165.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900173796.hg.1 | TC0900010850.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900173794.hg.1 | ENST00000416309.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900173794.hg.1 | ENST00000416309.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900173794.hg.1 | ENST00000602602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900173794.hg.1 | NR_046163.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900173794.hg.1 | NR_046165.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900173794.hg.1 | ENST00000416309.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900173794.hg.1 | ENST00000416309.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900173794.hg.1 | ENST00000602602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900173794.hg.1 | NR_046163.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900173794.hg.1 | NR_046165.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900173794.hg.1 | TC0900010850.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900173795.hg.1 | ENST00000416309.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900173795.hg.1 | ENST00000416309.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900173795.hg.1 | NR_046163.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900173795.hg.1 | ENST00000416309.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900173795.hg.1 | ENST00000416309.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900173795.hg.1 | NR_046163.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900173795.hg.1 | TC0900010850.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 1559129_a_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1559129_PM_a_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559129_a_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559129_a_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559129_a_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559129_a_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559129_a_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559129_a_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559129_a_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559129_a_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559129_a_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559129_a_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559129_a_at1 | - | - |
| Human Genome U95 Set | 71249_at | - | - |
| Human Genome U95 Set | 71249_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 229569 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1677834 | BC045813.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1677834 | BC036695.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1677834 | BC045813.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1677834 | BC036695.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1677834 | BC045813.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1677834 | BC036695.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1677834 | BC045813.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1677834 | BC036695.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1677834 | BC045813.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1677834 | BC036695.1 | human_transcriptome |

MIRLET7DHG
Genomic Map