Assay Details
| Gene Symbol: MUC5AC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001304359.1 | NP_001291288.1 | 3867 | 59 | ||
| GenBank mRNA | AF043909.1 | - | 29 - 30 | 3861 | 59 | |
| AJ298317.1 | - | 29 - 30 | 3858 | 59 | ||
| X81649.1 | - | 6 - 7 | 809 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

MUC5AC
Genomic Map