Assay Details
Gene Symbol: C17orf77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001302809.1 | NP_001289738.1 | 2 - 3 | 461 | 136 | |
NM_152460.2 | NP_689673.2 | 2 - 3 | 297 | 136 | ||
GenBank mRNA | AK056444.1 | - | 2 - 3 | 297 | 136 | |
BX957212.1 | - | 2 - 3 | 461 | 136 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17000832.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_001302809.1,NM_152460.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR17010750.hg.1 | ENST00000328023.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17010750.hg.1 | ENST00000392620.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17010750.hg.1 | NM_152460.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17010750.hg.1 | TC17000832.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR17010749.hg.1 | ENST00000328023.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17010749.hg.1 | ENST00000392620.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17010749.hg.1 | NM_152460.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17010749.hg.1 | TC17000832.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | ENST00000328023.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | ENST00000328023.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | ENST00000328023.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | ENST00000328023.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | ENST00000328023.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | ENST00000392620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | ENST00000392620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | ENST00000392620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | ENST00000392620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | ENST00000392620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | ENST00000392620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | ENST00000524389.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | ENST00000524389.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | ENST00000524389.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | ENST00000524389.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | NM_001302809.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | NM_001302809.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | NM_001302809.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | NM_001302809.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | NM_001302809.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | NM_001302809.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | NM_152460.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | NM_152460.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | NM_152460.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | NM_152460.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | NM_152460.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | NM_152460.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17010746.hg.1 | ENST00000392620.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17010746.hg.1 | NM_152460.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17010746.hg.1 | TC17000832.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1700011703.hg.1 | ENST00000375352.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011703.hg.1 | ENST00000375352.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011703.hg.1 | ENST00000375352.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011703.hg.1 | ENST00000375352.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011703.hg.1 | ENST00000375352.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011703.hg.1 | ENST00000375352.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011703.hg.1 | ENST00000375352.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011703.hg.1 | ENST00000375352.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011703.hg.1 | NM_001115152.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011703.hg.1 | NM_001115152.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011703.hg.1 | NM_001115152.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011703.hg.1 | NM_001115152.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011703.hg.1 | NM_001115152.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011703.hg.1 | NM_001115152.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17010748.hg.1 | ENST00000328023.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17010748.hg.1 | ENST00000392620.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17010748.hg.1 | NM_152460.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17010748.hg.1 | TC17000832.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0100018266.hg.1 | ENST00000431955.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018266.hg.1 | ENST00000431955.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018266.hg.1 | ENST00000431955.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018266.hg.1 | ENST00000431955.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700166151.hg.1 | ENST00000524389.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700166151.hg.1 | NM_001302809.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700166151.hg.1 | ENST00000524389.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700166151.hg.1 | NM_001302809.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700166151.hg.1 | TC1700008859.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1700011703.hg.1 | ENST00000375352.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011703.hg.1 | ENST00000375352.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011703.hg.1 | ENST00000375352.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011703.hg.1 | ENST00000375352.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011703.hg.1 | ENST00000375352.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011703.hg.1 | ENST00000375352.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011703.hg.1 | ENST00000375352.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011703.hg.1 | ENST00000375352.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011703.hg.1 | NM_001115152.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011703.hg.1 | NM_001115152.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011703.hg.1 | NM_001115152.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011703.hg.1 | NM_001115152.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011703.hg.1 | NM_001115152.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011703.hg.1 | NM_001115152.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700166154.hg.1 | ENST00000328023.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700166154.hg.1 | ENST00000392620.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700166154.hg.1 | ENST00000524389.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700166154.hg.1 | NM_001302809.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700166154.hg.1 | NM_152460.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700166154.hg.1 | ENST00000328023.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700166154.hg.1 | ENST00000392620.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700166154.hg.1 | ENST00000524389.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700166154.hg.1 | NM_001302809.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700166154.hg.1 | NM_152460.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700166154.hg.1 | TC1700008859.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700166156.hg.1 | ENST00000328023.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700166156.hg.1 | ENST00000392620.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700166156.hg.1 | ENST00000524389.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700166156.hg.1 | NM_001302809.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700166156.hg.1 | NM_152460.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700166156.hg.1 | ENST00000328023.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700166156.hg.1 | ENST00000392620.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700166156.hg.1 | ENST00000524389.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700166156.hg.1 | NM_001302809.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700166156.hg.1 | NM_152460.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700166156.hg.1 | TC1700008859.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700166157.hg.1 | ENST00000328023.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700166157.hg.1 | ENST00000392620.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700166157.hg.1 | NM_001302809.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700166157.hg.1 | NM_152460.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700166157.hg.1 | ENST00000328023.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700166157.hg.1 | ENST00000392620.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700166157.hg.1 | NM_001302809.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700166157.hg.1 | NM_152460.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700166157.hg.1 | TC1700008859.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | ENST00000328023.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | ENST00000328023.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | ENST00000328023.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | ENST00000328023.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | ENST00000328023.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | ENST00000392620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | ENST00000392620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | ENST00000392620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | ENST00000392620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | ENST00000392620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | ENST00000392620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | ENST00000524389.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | ENST00000524389.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | ENST00000524389.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | ENST00000524389.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | NM_001302809.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | NM_001302809.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | NM_001302809.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | NM_001302809.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | NM_001302809.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | NM_001302809.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | NM_152460.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | NM_152460.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | NM_152460.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | NM_152460.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | NM_152460.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | NM_152460.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018266.hg.1 | ENST00000431955.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018266.hg.1 | ENST00000431955.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018266.hg.1 | ENST00000431955.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018266.hg.1 | ENST00000431955.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700166152.hg.1 | ENST00000392620.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700166152.hg.1 | NM_152460.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700166152.hg.1 | ENST00000392620.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700166152.hg.1 | NM_152460.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700166152.hg.1 | TC1700008859.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700166155.hg.1 | ENST00000328023.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700166155.hg.1 | ENST00000392620.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700166155.hg.1 | ENST00000524389.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700166155.hg.1 | NM_001302809.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700166155.hg.1 | NM_152460.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700166155.hg.1 | ENST00000328023.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700166155.hg.1 | ENST00000392620.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700166155.hg.1 | ENST00000524389.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700166155.hg.1 | NM_001302809.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700166155.hg.1 | NM_152460.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700166155.hg.1 | TC1700008859.hg.1 | Clariom D TC |
