Assay Details
| Gene Symbol: SP2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_103857.1 | 2 - 3 | 183 | 61 | ||
| GenBank mRNA | BM925640.1 | - | 2 - 3 | 187 | 61 | |
| BX097765.1 | - | 2 - 3 | 147 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human 1B Microarray G4111A | A_32_P499570 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000411573.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000411573.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000411573.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000411573.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000411573.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000411573.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000411573.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000411573.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000411573.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000411573.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000433001.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000433001.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000433001.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000433001.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000585280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000585280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000585280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000585280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000585280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000585280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000585280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | ENST00000585280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | NR_103856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | NR_103856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | NR_103856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | NR_103856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | NR_103856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | NR_103856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | NR_103856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | NR_103856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | NR_103857.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | NR_103857.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | NR_103857.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | NR_103857.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012429.hg.1 | NR_103857.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000411573.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000411573.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000411573.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000411573.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000411573.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000411573.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000411573.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000411573.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000411573.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000411573.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000433001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000433001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000433001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000433001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000585280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000585280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000585280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000585280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000585280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000585280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000585280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | ENST00000585280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | NR_103856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | NR_103856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | NR_103856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | NR_103856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | NR_103856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | NR_103856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | NR_103856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | NR_103856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | NR_103857.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | NR_103857.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | NR_103857.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | NR_103857.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012429.hg.1 | NR_103857.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700206709.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206709.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206709.hg.1 | NR_103856.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700206709.hg.1 | NR_103857.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700206709.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206709.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206709.hg.1 | NR_103856.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700206709.hg.1 | NR_103857.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700206709.hg.1 | TC1700012429.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700206708.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206708.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206708.hg.1 | NR_103856.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700206708.hg.1 | NR_103857.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700206708.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206708.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206708.hg.1 | NR_103856.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700206708.hg.1 | NR_103857.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700206708.hg.1 | TC1700012429.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700206710.hg.1 | ENST00000411573.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206710.hg.1 | ENST00000411573.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206710.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206710.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206710.hg.1 | NR_103856.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700206710.hg.1 | NR_103857.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700206710.hg.1 | ENST00000411573.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206710.hg.1 | ENST00000411573.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206710.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206710.hg.1 | ENST00000451140.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206710.hg.1 | NR_103856.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700206710.hg.1 | NR_103857.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700206710.hg.1 | TC1700012429.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 232099 | - | - |

SP2-AS1
Genomic Map