Assay Details
| Gene Symbol: LINC01504 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110952.1 | 1 - 2 | 192 | 75 | ||
| NR_110953.1 | 1 - 2 | 192 | 75 | |||
| GenBank mRNA | AA436218.1 | - | 1 - 2 | 110 | 75 | |
| AK095210.1 | - | 1 - 2 | 192 | 75 | ||
More Information
Set Membership: |
|

LINC01504
Genomic Map