Assay Details
| Gene Symbol: C1QTNF1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040019.1 | 2 - 3 | 512 | 83 | ||
| GenBank mRNA | BC039683.1 | - | 2 - 3 | 458 | 83 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001926.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3354374 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P20739045 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P20739045 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P03296427 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 64895 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 32405 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P20739045 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03296422 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03296419 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03296427 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17026243.hg.1 | ENST00000397571.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17026243.hg.1 | NR_040018.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17026243.hg.1 | NR_040019.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17026243.hg.1 | TC17001926.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17026244.hg.1 | NR_040018.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17026244.hg.1 | NR_040019.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17026244.hg.1 | TC17001926.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17026240.hg.1 | ENST00000397571.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17026240.hg.1 | NR_040018.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17026240.hg.1 | NR_040019.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17026240.hg.1 | TC17001926.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17026242.hg.1 | ENST00000397571.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17026242.hg.1 | NR_040018.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17026242.hg.1 | NR_040019.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17026242.hg.1 | TC17001926.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011926.hg.1 | ENST00000577521.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011926.hg.1 | ENST00000577521.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011926.hg.1 | ENST00000577521.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011926.hg.1 | ENST00000577521.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011926.hg.1 | ENST00000577521.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011926.hg.1 | ENST00000581579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011926.hg.1 | ENST00000581579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011926.hg.1 | ENST00000581579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011926.hg.1 | ENST00000581579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011926.hg.1 | NR_040018.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011926.hg.1 | NR_040018.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011926.hg.1 | NR_040018.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011926.hg.1 | NR_040018.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011926.hg.1 | NR_040018.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011926.hg.1 | NR_040018.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011926.hg.1 | NR_040019.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011926.hg.1 | NR_040019.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011926.hg.1 | NR_040019.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011926.hg.1 | NR_040019.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011926.hg.1 | NR_040019.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197495.hg.1 | ENST00000577521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197495.hg.1 | ENST00000581579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197495.hg.1 | NR_040018.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197495.hg.1 | NR_040019.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197495.hg.1 | ENST00000577521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197495.hg.1 | ENST00000581579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197495.hg.1 | NR_040018.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197495.hg.1 | NR_040019.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197495.hg.1 | TC1700011926.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700011926.hg.1 | ENST00000577521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011926.hg.1 | ENST00000577521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011926.hg.1 | ENST00000577521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011926.hg.1 | ENST00000577521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011926.hg.1 | ENST00000577521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011926.hg.1 | ENST00000581579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011926.hg.1 | ENST00000581579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011926.hg.1 | ENST00000581579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011926.hg.1 | ENST00000581579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011926.hg.1 | NR_040018.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011926.hg.1 | NR_040018.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011926.hg.1 | NR_040018.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011926.hg.1 | NR_040018.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011926.hg.1 | NR_040018.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011926.hg.1 | NR_040018.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011926.hg.1 | NR_040019.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011926.hg.1 | NR_040019.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011926.hg.1 | NR_040019.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011926.hg.1 | NR_040019.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011926.hg.1 | NR_040019.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197498.hg.1 | ENST00000577521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197498.hg.1 | ENST00000581579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197498.hg.1 | NR_040018.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197498.hg.1 | NR_040019.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197498.hg.1 | ENST00000577521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197498.hg.1 | ENST00000581579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197498.hg.1 | NR_040018.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197498.hg.1 | NR_040019.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197498.hg.1 | TC1700011926.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700197499.hg.1 | ENST00000577521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197499.hg.1 | NR_040018.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197499.hg.1 | NR_040019.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197499.hg.1 | ENST00000577521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197499.hg.1 | NR_040018.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197499.hg.1 | NR_040019.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197499.hg.1 | TC1700011926.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700197497.hg.1 | ENST00000577521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197497.hg.1 | ENST00000581579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197497.hg.1 | NR_040018.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197497.hg.1 | NR_040019.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197497.hg.1 | ENST00000577521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197497.hg.1 | ENST00000581579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197497.hg.1 | NR_040018.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197497.hg.1 | NR_040019.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197497.hg.1 | TC1700011926.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A008854 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A008854 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina human-6 v2.0 expression beadchip | ILMN_1907318 | - | - |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 6270192 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_77369 | - | - |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1907318 | - | - |

C1QTNF1-AS1
Genomic Map