Assay Details
| Gene Symbol: LOC101927877 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135634.1 | 1 - 2 | 450 | 70 | ||
| GenBank mRNA | BC040294.1 | - | 1 - 2 | 452 | 70 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | membrane associated ring-CH-type finger 10 | MARCH10 |
MARCH-X, RNF190 |
-119.0 | Anti-sense | Chr. 17 - 62701314 - 62808381 on NCBI Build GRCh38 |
| Downstream | microRNA 633 | MIR633 |
MIRN633, hsa-mir-633 |
107.8kb | Sense | Chr. 17 - 62944215 - 62944312 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC101927877
Genomic Map