Assay Details
| Gene Symbol: LOC100130357 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242698.1 | NP_001229627.1 | 1 - 2 | 100 | 75 | |
| GenBank mRNA | AK091836.1 | - | 1 - 2 | 99 | 75 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001279.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_001242698.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P359067 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P359067 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P359067 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 1559507_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015330.hg.1 | TC06001279.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015330.hg.1 | NM_001242698.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014240.hg.1 | ENST00000399446.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014240.hg.1 | ENST00000399446.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014240.hg.1 | ENST00000399446.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014240.hg.1 | ENST00000399446.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014240.hg.1 | ENST00000399446.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014240.hg.1 | ENST00000399446.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014240.hg.1 | ENST00000606150.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014240.hg.1 | ENST00000606150.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014240.hg.1 | ENST00000606150.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014240.hg.1 | ENST00000606150.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014240.hg.1 | ENST00000606150.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014240.hg.1 | ENST00000606150.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014240.hg.1 | ENST00000606150.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014240.hg.1 | ENST00000606150.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014240.hg.1 | ENST00000606150.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014240.hg.1 | ENST00000606150.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014240.hg.1 | ENST00000606150.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014240.hg.1 | ENST00000606150.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014240.hg.1 | ENST00000606150.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014240.hg.1 | ENST00000606150.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014240.hg.1 | ENST00000606627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014240.hg.1 | ENST00000606627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014240.hg.1 | ENST00000606627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014240.hg.1 | ENST00000612479.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014240.hg.1 | ENST00000612479.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014240.hg.1 | ENST00000612479.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014240.hg.1 | ENST00000612479.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014240.hg.1 | NM_001242698.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014240.hg.1 | NM_001242698.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014240.hg.1 | NM_001242698.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014240.hg.1 | NM_001242698.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014240.hg.1 | NM_001242698.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014240.hg.1 | NM_001242698.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014240.hg.1 | NM_001242698.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015331.hg.1 | TC06001279.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015331.hg.1 | NM_001242698.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600200721.hg.1 | ENST00000606150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600200721.hg.1 | ENST00000606150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600200721.hg.1 | NM_001242698.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600200721.hg.1 | ENST00000606150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600200721.hg.1 | ENST00000606150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600200721.hg.1 | NM_001242698.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600200721.hg.1 | TC0600014240.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600200720.hg.1 | ENST00000606150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600200720.hg.1 | ENST00000606150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600200720.hg.1 | NM_001242698.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600200720.hg.1 | ENST00000606150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600200720.hg.1 | ENST00000606150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600200720.hg.1 | NM_001242698.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600200720.hg.1 | TC0600014240.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600014240.hg.1 | ENST00000399446.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014240.hg.1 | ENST00000399446.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014240.hg.1 | ENST00000399446.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014240.hg.1 | ENST00000399446.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014240.hg.1 | ENST00000399446.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014240.hg.1 | ENST00000399446.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014240.hg.1 | ENST00000606150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014240.hg.1 | ENST00000606150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014240.hg.1 | ENST00000606150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014240.hg.1 | ENST00000606150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014240.hg.1 | ENST00000606150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014240.hg.1 | ENST00000606150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014240.hg.1 | ENST00000606150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014240.hg.1 | ENST00000606150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014240.hg.1 | ENST00000606150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014240.hg.1 | ENST00000606150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014240.hg.1 | ENST00000606150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014240.hg.1 | ENST00000606150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014240.hg.1 | ENST00000606150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014240.hg.1 | ENST00000606150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014240.hg.1 | ENST00000606627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014240.hg.1 | ENST00000606627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014240.hg.1 | ENST00000606627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014240.hg.1 | ENST00000612479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014240.hg.1 | ENST00000612479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014240.hg.1 | ENST00000612479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014240.hg.1 | ENST00000612479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014240.hg.1 | NM_001242698.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014240.hg.1 | NM_001242698.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014240.hg.1 | NM_001242698.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014240.hg.1 | NM_001242698.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014240.hg.1 | NM_001242698.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014240.hg.1 | NM_001242698.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014240.hg.1 | NM_001242698.1 | Refseq |
| Human Genome U133 Plus 2.0 Array | 1559507_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1559507_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559507_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559507_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559507_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559507_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559507_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559507_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559507_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559507_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559507_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559507_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559507_at3 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A029423 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A029423 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3262298 | NM_001242698.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3262298 | AK091836.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3262298 | BX641137.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3262298 | AK296391.1 | human_transcriptome |

LOC100130357
Genomic Map