Assay Details
| Gene Symbol: CERS6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256126.1 | NP_001243055.1 | 5 - 6 | 714 | 78 | |
| NM_203463.2 | NP_982288.1 | 5 - 6 | 714 | 78 | ||
| XM_005246440.4 | XP_005246497.1 | 2 - 3 | 2005 | 78 | ||
| XM_017003749.1 | XP_016859238.1 | 2 - 3 | 865 | 78 | ||
| GenBank mRNA | AK172775.1 | - | 5 - 6 | 714 | 78 | |
| BC109284.1 | - | 5 - 6 | 564 | 40031838 | 78 | |
| BC109285.1 | - | 5 - 6 | 564 | 40031840 | 78 | |
| BE745491.1 | - | 231 | 78 | |||
| BG206657.1 | - | 2 - 3 | 120 | 78 | ||
| BX393696.2 | - | 5 - 6 | 644 | 78 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000996.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_005246440.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02014538.hg.1 | TC02000996.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02014538.hg.1 | ENST00000305747.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02014538.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02014538.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02014538.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02014537.hg.1 | TC02000996.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02014537.hg.1 | ENST00000305747.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02014537.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02014537.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02014537.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02014539.hg.1 | TC02000996.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02014539.hg.1 | ENST00000305747.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02014539.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02014539.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02014539.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169072.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169072.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169072.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169072.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169072.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169072.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169072.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169072.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169072.hg.1 | TC0200009895.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200169070.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169070.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169070.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169070.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169070.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169070.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169070.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169070.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169070.hg.1 | TC0200009895.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009895.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169071.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169071.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169071.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169071.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169071.hg.1 | ENST00000305747.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169071.hg.1 | ENST00000392687.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169071.hg.1 | NM_001256126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169071.hg.1 | NM_203463.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169071.hg.1 | TC0200009895.hg.1 | Clariom D TC |

CERS6
Genomic Map