Assay Details
| Gene Symbol: PAQR7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005245745.3 | XP_005245802.1 | 1 - 2 | 380 | 103 | |
| XM_011540862.2 | XP_011539164.1 | 1 - 2 | 382 | 103 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005245745.3, XM_011540862.2 transcripts.
Set Membership: |
|

PAQR7
Genomic Map