Assay Details
| Gene Symbol: KRT39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213656.3 | NP_998821.3 | 1 - 2 | 502 | 89 | |
| GenBank mRNA | AJ786657.1 | - | 1 - 2 | 502 | 89 | |
| BC157887.1 | - | 1 - 2 | 496 | 8862708 | 89 | |
| BC171907.1 | - | 1 - 2 | 496 | 89 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17002918.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17019457.hg.1 | TC17002918.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019457.hg.1 | ENST00000355612.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019457.hg.1 | NM_213656.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000355612.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000355612.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000355612.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000355612.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000355612.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000355612.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000355612.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000355612.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000355612.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000355612.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000355612.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000578029.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000578029.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000578029.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000578029.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000578078.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000578078.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000578078.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000578078.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000578078.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000578078.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000578078.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000578078.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000578078.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000578078.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | ENST00000578078.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | NM_213656.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | NM_213656.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | NM_213656.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | NM_213656.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | NM_213656.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | NM_213656.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | NM_213656.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | NM_213656.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | NM_213656.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | NM_213656.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012400.hg.1 | NM_213656.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019455.hg.1 | TC17002918.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019455.hg.1 | ENST00000355612.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019455.hg.1 | NM_213656.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019456.hg.1 | TC17002918.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019456.hg.1 | ENST00000355612.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019456.hg.1 | NM_213656.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700206295.hg.1 | ENST00000355612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206295.hg.1 | ENST00000578078.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206295.hg.1 | NM_213656.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700206295.hg.1 | ENST00000355612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206295.hg.1 | ENST00000578078.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206295.hg.1 | NM_213656.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700206295.hg.1 | TC1700012400.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700206296.hg.1 | ENST00000355612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206296.hg.1 | ENST00000578078.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206296.hg.1 | NM_213656.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700206296.hg.1 | ENST00000355612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206296.hg.1 | ENST00000578078.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206296.hg.1 | NM_213656.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700206296.hg.1 | TC1700012400.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700206294.hg.1 | ENST00000355612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206294.hg.1 | ENST00000578078.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206294.hg.1 | NM_213656.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700206294.hg.1 | ENST00000355612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206294.hg.1 | ENST00000578078.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206294.hg.1 | NM_213656.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700206294.hg.1 | TC1700012400.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000355612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000355612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000355612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000355612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000355612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000355612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000355612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000355612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000355612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000355612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000355612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000578029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000578029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000578029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000578029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000578078.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000578078.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000578078.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000578078.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000578078.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000578078.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000578078.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000578078.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000578078.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000578078.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | ENST00000578078.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | NM_213656.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | NM_213656.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | NM_213656.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | NM_213656.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | NM_213656.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | NM_213656.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | NM_213656.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | NM_213656.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | NM_213656.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | NM_213656.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012400.hg.1 | NM_213656.3 | Refseq |

KRT39
Genomic Map