Assay Details
Gene Symbol: C2orf83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162483.1 | NP_001155955.1 | 1 - 2 | 352 | 249 | |
NM_020161.3 | NP_064546.3 | 1 - 2 | 352 | 154 | ||
GenBank mRNA | AK055355.1 | - | 1 - 2 | 352 | 249 | |
AL359944.1 | - | 1 - 2 | 204 | 154 | ||
BC131618.1 | - | 1 - 2 | 190 | 40116809 | 154 | |
CR749679.1 | - | 1 - 2 | 204 | 154 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02002836.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR02045829.hg.1 | TC02002836.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02045829.hg.1 | ENST00000409066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02045829.hg.1 | NM_001162483.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02045830.hg.1 | TC02002836.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02045830.hg.1 | ENST00000264387.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02045830.hg.1 | ENST00000409066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02045830.hg.1 | NM_001162483.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02045830.hg.1 | NM_020161.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015929.hg.1 | ENST00000264387.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015929.hg.1 | ENST00000264387.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015929.hg.1 | ENST00000264387.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015929.hg.1 | ENST00000264387.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015929.hg.1 | ENST00000264387.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015929.hg.1 | ENST00000264387.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015929.hg.1 | ENST00000264387.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015929.hg.1 | ENST00000409066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015929.hg.1 | ENST00000409066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015929.hg.1 | ENST00000409066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015929.hg.1 | ENST00000409066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015929.hg.1 | ENST00000409066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015929.hg.1 | ENST00000409066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015929.hg.1 | ENST00000409066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015929.hg.1 | ENST00000409066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015929.hg.1 | ENST00000409066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015929.hg.1 | ENST00000509872.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015929.hg.1 | ENST00000509872.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015929.hg.1 | ENST00000509872.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015929.hg.1 | ENST00000509872.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015929.hg.1 | ENST00000509872.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015929.hg.1 | NM_001162483.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015929.hg.1 | NM_001162483.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015929.hg.1 | NM_001162483.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015929.hg.1 | NM_001162483.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015929.hg.1 | NM_001162483.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015929.hg.1 | NM_001162483.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015929.hg.1 | NM_001162483.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015929.hg.1 | NM_001162483.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015929.hg.1 | NM_001162483.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015929.hg.1 | NM_020161.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015929.hg.1 | NM_020161.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015929.hg.1 | NM_020161.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015929.hg.1 | NM_020161.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015929.hg.1 | NM_020161.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015929.hg.1 | NM_020161.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015929.hg.1 | NM_020161.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015929.hg.1 | NM_020161.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02045831.hg.1 | TC02002836.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02045831.hg.1 | ENST00000264387.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02045831.hg.1 | ENST00000409066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02045831.hg.1 | NM_001162483.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02045831.hg.1 | NM_020161.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02045833.hg.1 | TC02002836.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02045833.hg.1 | ENST00000409066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02045833.hg.1 | NM_001162483.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02045833.hg.1 | NM_020161.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200217223.hg.1 | ENST00000264387.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200217223.hg.1 | ENST00000409066.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200217223.hg.1 | NM_001162483.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200217223.hg.1 | NM_020161.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200217223.hg.1 | ENST00000264387.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200217223.hg.1 | ENST00000409066.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200217223.hg.1 | NM_001162483.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200217223.hg.1 | NM_020161.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200217223.hg.1 | TC0200015929.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200217225.hg.1 | ENST00000264387.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200217225.hg.1 | ENST00000409066.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200217225.hg.1 | ENST00000509872.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200217225.hg.1 | NM_001162483.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200217225.hg.1 | NM_020161.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200217225.hg.1 | ENST00000264387.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200217225.hg.1 | ENST00000409066.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200217225.hg.1 | ENST00000509872.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200217225.hg.1 | NM_001162483.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200217225.hg.1 | NM_020161.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200217225.hg.1 | TC0200015929.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200217224.hg.1 | ENST00000264387.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200217224.hg.1 | ENST00000409066.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200217224.hg.1 | ENST00000509872.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200217224.hg.1 | NM_001162483.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200217224.hg.1 | NM_020161.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200217224.hg.1 | ENST00000264387.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200217224.hg.1 | ENST00000409066.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200217224.hg.1 | ENST00000509872.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200217224.hg.1 | NM_001162483.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200217224.hg.1 | NM_020161.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200217224.hg.1 | TC0200015929.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200217228.hg.1 | ENST00000409066.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200217228.hg.1 | NM_001162483.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200217228.hg.1 | NM_020161.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200217228.hg.1 | ENST00000409066.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200217228.hg.1 | NM_001162483.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200217228.hg.1 | NM_020161.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200217228.hg.1 | TC0200015929.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200217222.hg.1 | ENST00000409066.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200217222.hg.1 | NM_001162483.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200217222.hg.1 | ENST00000409066.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200217222.hg.1 | NM_001162483.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200217222.hg.1 | TC0200015929.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200217226.hg.1 | ENST00000264387.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200217226.hg.1 | ENST00000409066.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200217226.hg.1 | NM_001162483.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200217226.hg.1 | NM_020161.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200217226.hg.1 | ENST00000264387.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200217226.hg.1 | ENST00000409066.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200217226.hg.1 | NM_001162483.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200217226.hg.1 | NM_020161.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200217226.hg.1 | TC0200015929.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0200015929.hg.1 | ENST00000264387.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015929.hg.1 | ENST00000264387.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015929.hg.1 | ENST00000264387.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015929.hg.1 | ENST00000264387.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015929.hg.1 | ENST00000264387.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015929.hg.1 | ENST00000264387.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015929.hg.1 | ENST00000264387.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015929.hg.1 | ENST00000409066.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015929.hg.1 | ENST00000409066.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015929.hg.1 | ENST00000409066.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015929.hg.1 | ENST00000409066.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015929.hg.1 | ENST00000409066.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015929.hg.1 | ENST00000409066.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015929.hg.1 | ENST00000409066.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015929.hg.1 | ENST00000409066.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015929.hg.1 | ENST00000409066.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015929.hg.1 | ENST00000509872.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015929.hg.1 | ENST00000509872.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015929.hg.1 | ENST00000509872.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015929.hg.1 | ENST00000509872.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015929.hg.1 | ENST00000509872.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015929.hg.1 | NM_001162483.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015929.hg.1 | NM_001162483.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015929.hg.1 | NM_001162483.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015929.hg.1 | NM_001162483.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015929.hg.1 | NM_001162483.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015929.hg.1 | NM_001162483.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015929.hg.1 | NM_001162483.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015929.hg.1 | NM_001162483.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015929.hg.1 | NM_001162483.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015929.hg.1 | NM_020161.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015929.hg.1 | NM_020161.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015929.hg.1 | NM_020161.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015929.hg.1 | NM_020161.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015929.hg.1 | NM_020161.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015929.hg.1 | NM_020161.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015929.hg.1 | NM_020161.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015929.hg.1 | NM_020161.3 | Refseq |
