Assay Details
Gene Symbol: ARL5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143968.1 | NP_001137440.1 | 1 - 2 | 449 | 65 | |
GenBank mRNA | AK310706.1 | - | 3 - 4 | 388 | 65 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17001445.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1700010562.hg.1 | ENST00000269586.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010562.hg.1 | ENST00000269586.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010562.hg.1 | ENST00000269586.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010562.hg.1 | ENST00000269586.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010562.hg.1 | ENST00000269586.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010562.hg.1 | ENST00000269586.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010562.hg.1 | ENST00000578912.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010562.hg.1 | ENST00000578912.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010562.hg.1 | ENST00000578912.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010562.hg.1 | ENST00000578912.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010562.hg.1 | ENST00000581255.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010562.hg.1 | ENST00000581255.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010562.hg.1 | ENST00000581255.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010562.hg.1 | ENST00000581255.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010562.hg.1 | ENST00000581255.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010562.hg.1 | ENST00000581554.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010562.hg.1 | ENST00000581554.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010562.hg.1 | ENST00000581554.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010562.hg.1 | ENST00000581554.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010562.hg.1 | ENST00000581554.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010562.hg.1 | ENST00000581554.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010562.hg.1 | ENST00000581554.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010562.hg.1 | ENST00000581554.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010562.hg.1 | ENST00000581554.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010562.hg.1 | ENST00000581554.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010562.hg.1 | ENST00000583123.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010562.hg.1 | ENST00000583123.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010562.hg.1 | ENST00000583123.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010562.hg.1 | ENST00000583123.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010562.hg.1 | ENST00000583123.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010562.hg.1 | ENST00000583123.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010562.hg.1 | NM_001143968.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010562.hg.1 | NM_001143968.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010562.hg.1 | NM_001143968.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010562.hg.1 | NM_001143968.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010562.hg.1 | NM_001143968.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010562.hg.1 | NM_001143968.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010562.hg.1 | NM_001143968.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010562.hg.1 | NM_001143968.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17019046.hg.1 | ENST00000269586.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17019046.hg.1 | ENST00000444555.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17019046.hg.1 | NM_001143968.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17019046.hg.1 | TC17001445.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR17019047.hg.1 | ENST00000269586.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17019047.hg.1 | ENST00000444555.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17019047.hg.1 | NM_001143968.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17019047.hg.1 | TC17001445.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR17019048.hg.1 | ENST00000269586.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17019048.hg.1 | NM_001143968.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17019048.hg.1 | TC17001445.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1700183197.hg.1 | ENST00000269586.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700183197.hg.1 | ENST00000581255.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700183197.hg.1 | ENST00000581554.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700183197.hg.1 | ENST00000583123.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700183197.hg.1 | NM_001143968.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700183197.hg.1 | ENST00000269586.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700183197.hg.1 | ENST00000581255.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700183197.hg.1 | ENST00000581554.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700183197.hg.1 | ENST00000583123.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700183197.hg.1 | NM_001143968.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700183197.hg.1 | TC1700010562.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1700010562.hg.1 | ENST00000269586.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010562.hg.1 | ENST00000269586.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010562.hg.1 | ENST00000269586.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010562.hg.1 | ENST00000269586.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010562.hg.1 | ENST00000269586.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010562.hg.1 | ENST00000269586.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010562.hg.1 | ENST00000578912.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010562.hg.1 | ENST00000578912.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010562.hg.1 | ENST00000578912.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010562.hg.1 | ENST00000578912.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010562.hg.1 | ENST00000581255.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010562.hg.1 | ENST00000581255.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010562.hg.1 | ENST00000581255.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010562.hg.1 | ENST00000581255.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010562.hg.1 | ENST00000581255.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010562.hg.1 | ENST00000581554.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010562.hg.1 | ENST00000581554.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010562.hg.1 | ENST00000581554.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010562.hg.1 | ENST00000581554.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010562.hg.1 | ENST00000581554.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010562.hg.1 | ENST00000581554.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010562.hg.1 | ENST00000581554.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010562.hg.1 | ENST00000581554.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010562.hg.1 | ENST00000581554.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010562.hg.1 | ENST00000581554.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010562.hg.1 | ENST00000583123.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010562.hg.1 | ENST00000583123.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010562.hg.1 | ENST00000583123.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010562.hg.1 | ENST00000583123.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010562.hg.1 | ENST00000583123.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010562.hg.1 | ENST00000583123.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010562.hg.1 | NM_001143968.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010562.hg.1 | NM_001143968.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010562.hg.1 | NM_001143968.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010562.hg.1 | NM_001143968.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010562.hg.1 | NM_001143968.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010562.hg.1 | NM_001143968.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010562.hg.1 | NM_001143968.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010562.hg.1 | NM_001143968.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700183199.hg.1 | ENST00000581255.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700183199.hg.1 | ENST00000581554.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700183199.hg.1 | NM_001143968.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700183199.hg.1 | ENST00000581255.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700183199.hg.1 | ENST00000581554.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700183199.hg.1 | NM_001143968.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700183199.hg.1 | TC1700010562.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700183198.hg.1 | ENST00000581255.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700183198.hg.1 | ENST00000581554.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700183198.hg.1 | ENST00000583123.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700183198.hg.1 | NM_001143968.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700183198.hg.1 | ENST00000581255.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700183198.hg.1 | ENST00000581554.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700183198.hg.1 | ENST00000583123.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700183198.hg.1 | NM_001143968.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700183198.hg.1 | TC1700010562.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700183195.hg.1 | ENST00000269586.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700183195.hg.1 | ENST00000581255.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700183195.hg.1 | ENST00000581554.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700183195.hg.1 | ENST00000583123.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700183195.hg.1 | NM_001143968.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700183195.hg.1 | ENST00000269586.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700183195.hg.1 | ENST00000581255.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700183195.hg.1 | ENST00000581554.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700183195.hg.1 | ENST00000583123.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700183195.hg.1 | NM_001143968.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700183195.hg.1 | TC1700010562.hg.1 | Clariom D TC |
