Assay Details
| Gene Symbol: LOC101926960 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104636.1 | 3 - 4 | 327 | 96 | ||
| GenBank mRNA | AL691691.1 | - | 3 - 4 | 327 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

LOC101926960
Genomic Map