Assay Details
| Gene Symbol: LOC440934 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_136639.1 | 1 - 2 | 603 | 146 | ||
| GenBank mRNA | BC033986.1 | - | 1 - 2 | 569 | 146 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200010895.hg.1 | ENST00000424010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010895.hg.1 | ENST00000424010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010895.hg.1 | ENST00000424010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010895.hg.1 | ENST00000424010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010895.hg.1 | ENST00000429475.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010895.hg.1 | ENST00000429475.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010895.hg.1 | ENST00000429475.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010895.hg.1 | ENST00000429475.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010895.hg.1 | ENST00000429475.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010895.hg.1 | ENST00000429475.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010895.hg.1 | ENST00000429475.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010895.hg.1 | ENST00000429475.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010895.hg.1 | ENST00000429475.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010895.hg.1 | ENST00000439237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010895.hg.1 | ENST00000439237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010895.hg.1 | ENST00000439237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010895.hg.1 | ENST00000439237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010895.hg.1 | ENST00000446532.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010895.hg.1 | ENST00000446532.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010895.hg.1 | ENST00000446532.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010895.hg.1 | ENST00000446532.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010895.hg.1 | ENST00000446532.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010895.hg.1 | ENST00000555502.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010895.hg.1 | ENST00000555502.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010895.hg.1 | ENST00000555502.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010895.hg.1 | ENST00000555502.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010895.hg.1 | ENST00000555502.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010895.hg.1 | ENST00000555502.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010895.hg.1 | ENST00000555502.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010895.hg.1 | ENST00000555502.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010895.hg.1 | ENST00000587192.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010895.hg.1 | ENST00000587192.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010895.hg.1 | ENST00000587192.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010895.hg.1 | ENST00000587192.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010895.hg.1 | ENST00000587192.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010895.hg.1 | ENST00000587192.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010895.hg.1 | ENST00000589724.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010895.hg.1 | ENST00000589724.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010895.hg.1 | ENST00000589724.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010895.hg.1 | ENST00000589724.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010895.hg.1 | ENST00000590427.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010895.hg.1 | ENST00000590427.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010895.hg.1 | ENST00000590427.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010895.hg.1 | ENST00000424010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010895.hg.1 | ENST00000424010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010895.hg.1 | ENST00000424010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010895.hg.1 | ENST00000424010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010895.hg.1 | ENST00000429475.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010895.hg.1 | ENST00000429475.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010895.hg.1 | ENST00000429475.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010895.hg.1 | ENST00000429475.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010895.hg.1 | ENST00000429475.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010895.hg.1 | ENST00000429475.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010895.hg.1 | ENST00000429475.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010895.hg.1 | ENST00000429475.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010895.hg.1 | ENST00000429475.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010895.hg.1 | ENST00000439237.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010895.hg.1 | ENST00000439237.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010895.hg.1 | ENST00000439237.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010895.hg.1 | ENST00000439237.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010895.hg.1 | ENST00000446532.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010895.hg.1 | ENST00000446532.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010895.hg.1 | ENST00000446532.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010895.hg.1 | ENST00000446532.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010895.hg.1 | ENST00000446532.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010895.hg.1 | ENST00000555502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010895.hg.1 | ENST00000555502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010895.hg.1 | ENST00000555502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010895.hg.1 | ENST00000555502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010895.hg.1 | ENST00000555502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010895.hg.1 | ENST00000555502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010895.hg.1 | ENST00000555502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010895.hg.1 | ENST00000555502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010895.hg.1 | ENST00000587192.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010895.hg.1 | ENST00000587192.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010895.hg.1 | ENST00000587192.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010895.hg.1 | ENST00000587192.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010895.hg.1 | ENST00000587192.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010895.hg.1 | ENST00000587192.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010895.hg.1 | ENST00000589724.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010895.hg.1 | ENST00000589724.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010895.hg.1 | ENST00000589724.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010895.hg.1 | ENST00000589724.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010895.hg.1 | ENST00000590427.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010895.hg.1 | ENST00000590427.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010895.hg.1 | ENST00000590427.1 | Ensembl |

LOC440934
Genomic Map