Assay Details
| Gene Symbol: CATSPER4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198137.1 | NP_937770.1 | 5 - 6 | 683 | 77 | |
| XM_011541432.2 | XP_011539734.1 | 5 - 6 | 790 | 77 | ||
| XM_011541433.2 | XP_011539735.1 | 5 - 6 | 1349 | 77 | ||
| GenBank mRNA | BC128139.1 | - | 5 - 6 | 683 | 40112493 | 77 |
| BC144663.1 | - | 4 - 5 | 652 | 77 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000337.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
multicellular organism development
neuronal action potential
sperm motility
response to progesterone
regulation of ion transmembrane transport
sperm-egg recognition
sodium ion transmembrane transport
sperm capacitation
regulation of postsynaptic membrane potential
calcium ion transmembrane transport
membrane depolarization during action potential
calcium activated cation channel activity
voltage-gated calcium channel activity
voltage-gated sodium channel activity
multicellular organism development
neuronal action potential
sperm motility
response to progesterone
regulation of ion transmembrane transport
sperm-egg recognition
sodium ion transmembrane transport
sperm capacitation
regulation of postsynaptic membrane potential
calcium ion transmembrane transport
membrane depolarization during action potential
calcium activated cation channel activity
voltage-gated calcium channel activity
voltage-gated sodium channel activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01005171.hg.1 | TC01000337.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01005171.hg.1 | ENST00000456354.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01005171.hg.1 | NM_198137.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000338855.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000338855.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000338855.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000338855.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000338855.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000338855.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000338855.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000338855.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000338855.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000338855.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000338855.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000338855.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000518899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000518899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000518899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000518899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000518899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000518899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000518899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000518899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000518899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000518899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000518899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000518899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000611853.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000611853.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000611853.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000611853.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000611853.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000611853.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000611853.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000611853.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000611853.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000611853.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | ENST00000611853.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | NM_198137.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | NM_198137.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | NM_198137.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | NM_198137.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | NM_198137.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | NM_198137.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | NM_198137.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | NM_198137.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | NM_198137.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | NM_198137.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007445.hg.1 | NM_198137.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01005173.hg.1 | TC01000337.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01005173.hg.1 | ENST00000456354.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01005173.hg.1 | NM_198137.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01005172.hg.1 | TC01000337.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01005172.hg.1 | ENST00000456354.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01005172.hg.1 | NM_198137.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100151941.hg.1 | ENST00000338855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151941.hg.1 | ENST00000456354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151941.hg.1 | NM_198137.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100151941.hg.1 | ENST00000338855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151941.hg.1 | ENST00000456354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151941.hg.1 | NM_198137.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100151941.hg.1 | TC0100007445.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000338855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000338855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000338855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000338855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000338855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000338855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000338855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000338855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000338855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000338855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000338855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000338855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000456354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000518899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000518899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000518899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000518899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000518899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000518899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000518899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000518899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000518899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000518899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000518899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000518899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000611853.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000611853.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000611853.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000611853.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000611853.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000611853.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000611853.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000611853.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000611853.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000611853.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | ENST00000611853.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | NM_198137.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | NM_198137.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | NM_198137.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | NM_198137.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | NM_198137.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | NM_198137.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | NM_198137.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | NM_198137.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | NM_198137.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | NM_198137.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007445.hg.1 | NM_198137.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100151942.hg.1 | ENST00000338855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151942.hg.1 | ENST00000456354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151942.hg.1 | ENST00000518899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151942.hg.1 | ENST00000611853.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151942.hg.1 | NM_198137.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100151942.hg.1 | ENST00000338855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151942.hg.1 | ENST00000456354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151942.hg.1 | ENST00000518899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151942.hg.1 | ENST00000611853.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151942.hg.1 | NM_198137.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100151942.hg.1 | TC0100007445.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100151943.hg.1 | ENST00000338855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151943.hg.1 | ENST00000456354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151943.hg.1 | ENST00000518899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151943.hg.1 | ENST00000611853.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151943.hg.1 | NM_198137.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100151943.hg.1 | ENST00000338855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151943.hg.1 | ENST00000456354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151943.hg.1 | ENST00000518899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151943.hg.1 | ENST00000611853.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151943.hg.1 | NM_198137.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100151943.hg.1 | TC0100007445.hg.1 | Clariom D TC |

CATSPER4
Genomic Map