Assay Details
Gene Symbol: C12orf80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242696.1 | NP_001229625.1 | 3 - 4 | 690 | 62 | |
GenBank mRNA | BC038743.1 | - | 3 - 4 | 658 | 62 | |
BC117348.1 | - | 3 - 4 | 540 | 62 | ||
BC126393.1 | - | 3 - 4 | 540 | 62 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12001514.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08353085 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08353104 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08353094 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08353097 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08353105 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08353079 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08353068 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28787847 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08353082 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08353099 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08353125 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08353078 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08353127 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28787840 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08353124 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28787842 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08353123 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08353121 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28787880 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08353093 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08353115 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08353117 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08353074 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08353112 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28787878 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28787860 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08353077 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28787894 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08353114 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28787869 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08353067 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28787871 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08353110 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08353091 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28787855 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08353089 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08353095 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08353072 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08353129 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08353120 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08353076 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28787841 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28787856 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16558355 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28787885 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08353107 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16558360 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 25945 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 582218 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 633076 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 113881 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3606685 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 7733 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P02620609 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3606685 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3606685 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P02620609 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P02620609 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 165846 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 104603 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02620613 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02620609 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02620603 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P350664 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P02620609 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT HG-U133+ PM Array Plate | 1560277_PM_a_at | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 1560276_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR12019885.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12019885.hg.1 | NM_001242696.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12019885.hg.1 | TC12001514.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1200010741.hg.1 | ENST00000551332.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010741.hg.1 | ENST00000551332.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010741.hg.1 | ENST00000551332.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010741.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010741.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010741.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010741.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010741.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010741.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010741.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010741.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010741.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010741.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010741.hg.1 | ENST00000553017.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010741.hg.1 | ENST00000553017.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010741.hg.1 | ENST00000553017.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010741.hg.1 | NM_001242696.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010741.hg.1 | NM_001242696.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010741.hg.1 | NM_001242696.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010741.hg.1 | NM_001242696.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010741.hg.1 | NM_001242696.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010741.hg.1 | NM_001242696.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200181808.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200181808.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200181808.hg.1 | NM_001242696.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200181808.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200181808.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200181808.hg.1 | NM_001242696.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200181808.hg.1 | TC1200010741.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1200010741.hg.1 | ENST00000551332.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010741.hg.1 | ENST00000551332.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010741.hg.1 | ENST00000551332.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010741.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010741.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010741.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010741.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010741.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010741.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010741.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010741.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010741.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010741.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010741.hg.1 | ENST00000553017.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010741.hg.1 | ENST00000553017.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010741.hg.1 | ENST00000553017.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010741.hg.1 | NM_001242696.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010741.hg.1 | NM_001242696.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010741.hg.1 | NM_001242696.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010741.hg.1 | NM_001242696.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010741.hg.1 | NM_001242696.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010741.hg.1 | NM_001242696.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200181809.hg.1 | ENST00000551332.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200181809.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200181809.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200181809.hg.1 | NM_001242696.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200181809.hg.1 | ENST00000551332.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200181809.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200181809.hg.1 | ENST00000551894.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200181809.hg.1 | NM_001242696.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200181809.hg.1 | TC1200010741.hg.1 | Clariom D TC |
Human Genome U133 Plus 2.0 Array | 1560276_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 1560277_PM_a_at | - | - |
Human Genome U133 Plus 2.0 Array | 1560276_at | - | - |
Human Genome U133 Plus 2.0 Array | 1560277_a_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560276_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560277_a_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560277_a_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560277_a_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560276_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560276_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560276_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560276_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560277_a_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560277_a_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560276_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560277_a_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560277_a_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560276_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560276_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560276_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560277_a_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560277_a_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560276_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560277_a_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560276_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560277_a_at10 | - | - |
Human Genome U95 Set | 51262_at | - | - |
Human Genome U95 Set | 51262_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 700526 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina human-6 v2.0 expression beadchip | ILMN_1859233 | - | - |
Illumina Sentrix Human-6 v2 Expression BeadChip | 5490014 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_77924 | - | - |
Illumina HumanWG-6_V2_0_R2 | ILMN_1859233 | - | - |
