Assay Details
Gene Symbol: FAAHP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045483.1 | 5 - 6 | 468 | 76 | ||
GenBank mRNA | BI756183.1 | - | 2 - 3 | 159 | 76 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01000586.hg.1
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01010021.hg.1 | TC01000586.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01010021.hg.1 | ENST00000429784.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01010021.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01010020.hg.1 | TC01000586.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01010020.hg.1 | ENST00000429784.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01010020.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000429784.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000429784.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000429784.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000429784.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000429784.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000429784.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000429784.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000606395.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000606395.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000606395.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000606395.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000606395.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000607656.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000607656.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000607656.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000607656.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100158423.hg.1 | ENST00000429784.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158423.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100158423.hg.1 | ENST00000429784.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158423.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100158423.hg.1 | TC0100008160.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100158426.hg.1 | ENST00000429784.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158426.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158426.hg.1 | ENST00000607656.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158426.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100158426.hg.1 | ENST00000429784.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158426.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158426.hg.1 | ENST00000607656.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158426.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100158426.hg.1 | TC0100008160.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000429784.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000429784.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000429784.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000429784.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000429784.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000429784.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000429784.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000606395.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000606395.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000606395.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000606395.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000606395.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000607656.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000607656.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000607656.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000607656.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | NR_045483.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | NR_045483.1 | Refseq |
