Assay Details
| Gene Symbol: NBEAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114132.1 | NP_001107604.1 | 16 - 17 | 79 | ||
| XM_005246787.3 | XP_005246844.1 | 16 - 17 | 79 | |||
| XM_006712698.3 | XP_006712761.1 | 17 - 18 | 79 | |||
| XM_006712699.3 | XP_006712762.1 | 17 - 18 | 79 | |||
| XM_011511658.2 | XP_011509960.1 | 17 - 18 | 79 | |||
| XM_011511659.2 | XP_011509961.1 | 17 - 18 | 79 | |||
| XM_011511660.2 | XP_011509962.1 | 13 - 14 | 79 | |||
| XM_011511662.2 | XP_011509964.1 | 9 - 10 | 79 | |||
| XM_011511663.2 | XP_011509965.1 | 17 - 18 | 79 | |||
| XM_017004728.1 | XP_016860217.1 | 4 - 5 | 79 | |||
| GenBank mRNA | DV080456.1 | - | 4 - 5 | 456 | 79 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001195.hg.1
Set Membership: |
|

NBEAL1
Genomic Map