Assay Details
| Gene Symbol: PLEKHG4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006714446.3 | XP_006714509.1 | 2 - 3 | 360 | 76 | |
| XM_011513968.2 | XP_011512270.1 | 2 - 3 | 360 | 76 | ||
| XM_011513971.2 | XP_011512273.1 | 2 - 3 | 305 | 76 | ||
| XM_011513972.2 | XP_011512274.1 | 2 - 3 | 361 | 76 | ||
| XM_011513973.1 | XP_011512275.1 | 2 - 3 | 358 | 76 | ||
| XM_017009112.1 | XP_016864601.1 | 2 - 3 | 361 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011513971.2 transcripts.
Set Membership: |
|

PLEKHG4B
Genomic Map