Assay Details
| Gene Symbol: GPATCH11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278505.1 | NP_001265434.1 | 2 - 3 | 195 | 94 | |
| GenBank mRNA | AK095667.1 | - | 2 - 3 | 128 | 94 | |
| AK289504.1 | - | 2 - 3 | 110 | 94 | ||
| BC071798.1 | - | 2 - 3 | 127 | 94 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK095667.1,AK289504.1,NM_001278505.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000473067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000473067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000473067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000473067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000473067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200150158.hg.1 | ENST00000281932.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150158.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150158.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200150158.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200150158.hg.1 | ENST00000281932.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150158.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150158.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200150158.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200150158.hg.1 | TC0200007249.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200150157.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150157.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200150157.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200150157.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150157.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200150157.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200150157.hg.1 | TC0200007249.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000473067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000473067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000473067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000473067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000473067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200150160.hg.1 | ENST00000281932.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150160.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150160.hg.1 | ENST00000473067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150160.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200150160.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200150160.hg.1 | ENST00000281932.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150160.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150160.hg.1 | ENST00000473067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150160.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200150160.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200150160.hg.1 | TC0200007249.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200150164.hg.1 | ENST00000281932.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150164.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150164.hg.1 | ENST00000473067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150164.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200150164.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200150164.hg.1 | ENST00000281932.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150164.hg.1 | ENST00000409774.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150164.hg.1 | ENST00000473067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150164.hg.1 | NM_001278505.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200150164.hg.1 | NM_174931.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200150164.hg.1 | TC0200007249.hg.1 | Clariom D TC |

GPATCH11
Genomic Map