| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02003459.hg.1 |
TC02000233.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02003459.hg.1 |
ENST00000281932.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02003459.hg.1 |
ENST00000409774.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02003459.hg.1 |
NM_174931.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007249.hg.1 |
ENST00000281932.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007249.hg.1 |
ENST00000281932.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007249.hg.1 |
ENST00000281932.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007249.hg.1 |
ENST00000281932.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007249.hg.1 |
ENST00000281932.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007249.hg.1 |
ENST00000281932.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007249.hg.1 |
ENST00000281932.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007249.hg.1 |
ENST00000281932.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007249.hg.1 |
ENST00000281932.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007249.hg.1 |
ENST00000409774.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007249.hg.1 |
ENST00000409774.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007249.hg.1 |
ENST00000409774.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007249.hg.1 |
ENST00000409774.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007249.hg.1 |
ENST00000409774.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007249.hg.1 |
ENST00000409774.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007249.hg.1 |
ENST00000409774.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007249.hg.1 |
ENST00000409774.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007249.hg.1 |
ENST00000409774.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007249.hg.1 |
ENST00000409774.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007249.hg.1 |
ENST00000409774.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007249.hg.1 |
ENST00000409774.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007249.hg.1 |
ENST00000473067.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007249.hg.1 |
ENST00000473067.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007249.hg.1 |
ENST00000473067.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007249.hg.1 |
ENST00000473067.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007249.hg.1 |
ENST00000473067.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007249.hg.1 |
NM_001278505.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007249.hg.1 |
NM_001278505.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007249.hg.1 |
NM_001278505.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007249.hg.1 |
NM_001278505.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007249.hg.1 |
NM_001278505.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007249.hg.1 |
NM_001278505.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007249.hg.1 |
NM_001278505.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007249.hg.1 |
NM_001278505.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007249.hg.1 |
NM_001278505.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007249.hg.1 |
NM_001278505.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007249.hg.1 |
NM_174931.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007249.hg.1 |
NM_174931.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007249.hg.1 |
NM_174931.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007249.hg.1 |
NM_174931.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007249.hg.1 |
NM_174931.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007249.hg.1 |
NM_174931.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007249.hg.1 |
NM_174931.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007249.hg.1 |
NM_174931.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007249.hg.1 |
NM_174931.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007249.hg.1 |
NM_174931.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007249.hg.1 |
NM_174931.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007249.hg.1 |
NM_174931.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200150168.hg.1 |
ENST00000281932.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200150168.hg.1 |
ENST00000409774.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200150168.hg.1 |
NM_001278505.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200150168.hg.1 |
NM_174931.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200150168.hg.1 |
ENST00000281932.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200150168.hg.1 |
ENST00000409774.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200150168.hg.1 |
NM_001278505.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200150168.hg.1 |
NM_174931.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200150168.hg.1 |
TC0200007249.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200007249.hg.1 |
ENST00000281932.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007249.hg.1 |
ENST00000281932.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007249.hg.1 |
ENST00000281932.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007249.hg.1 |
ENST00000281932.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007249.hg.1 |
ENST00000281932.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007249.hg.1 |
ENST00000281932.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007249.hg.1 |
ENST00000281932.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007249.hg.1 |
ENST00000281932.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007249.hg.1 |
ENST00000281932.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007249.hg.1 |
ENST00000409774.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007249.hg.1 |
ENST00000409774.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007249.hg.1 |
ENST00000409774.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007249.hg.1 |
ENST00000409774.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007249.hg.1 |
ENST00000409774.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007249.hg.1 |
ENST00000409774.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007249.hg.1 |
ENST00000409774.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007249.hg.1 |
ENST00000409774.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007249.hg.1 |
ENST00000409774.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007249.hg.1 |
ENST00000409774.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007249.hg.1 |
ENST00000409774.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007249.hg.1 |
ENST00000409774.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007249.hg.1 |
ENST00000473067.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007249.hg.1 |
ENST00000473067.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007249.hg.1 |
ENST00000473067.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007249.hg.1 |
ENST00000473067.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007249.hg.1 |
ENST00000473067.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007249.hg.1 |
NM_001278505.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007249.hg.1 |
NM_001278505.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007249.hg.1 |
NM_001278505.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007249.hg.1 |
NM_001278505.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007249.hg.1 |
NM_001278505.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007249.hg.1 |
NM_001278505.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007249.hg.1 |
NM_001278505.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007249.hg.1 |
NM_001278505.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007249.hg.1 |
NM_001278505.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007249.hg.1 |
NM_001278505.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007249.hg.1 |
NM_174931.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007249.hg.1 |
NM_174931.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007249.hg.1 |
NM_174931.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007249.hg.1 |
NM_174931.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007249.hg.1 |
NM_174931.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007249.hg.1 |
NM_174931.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007249.hg.1 |
NM_174931.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007249.hg.1 |
NM_174931.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007249.hg.1 |
NM_174931.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007249.hg.1 |
NM_174931.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007249.hg.1 |
NM_174931.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007249.hg.1 |
NM_174931.3 |
Refseq |