Assay Details
| Gene Symbol: REG3G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008387.2 | NP_001008388.1 | 1 - 2 | 93 | 94 | |
| NM_001270040.1 | NP_001256969.1 | 1 - 2 | 93 | 94 | ||
| GenBank mRNA | AB161038.1 | - | 1 - 2 | 78 | 94 | |
| BC103852.2 | - | 1 - 2 | 52 | 40002928 | 94 | |
| BC103853.1 | - | 1 - 2 | 52 | 40002929 | 94 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000487.hg.1
Additional Information:
This assay is in the 5'UTR region of the AB161038.1,BC103852.2,BC103853.1,NM_001008387.2,NM_001270040.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
MyD88-dependent toll-like receptor signaling pathway
acute-phase response
positive regulation of keratinocyte proliferation
negative regulation of keratinocyte differentiation
defense response to Gram-positive bacterium
positive regulation of wound healing
carbohydrate binding
MyD88-dependent toll-like receptor signaling pathway
acute-phase response
positive regulation of keratinocyte proliferation
negative regulation of keratinocyte differentiation
defense response to Gram-positive bacterium
positive regulation of wound healing
carbohydrate binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02007457.hg.1 | TC02000487.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007457.hg.1 | ENST00000272324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007457.hg.1 | ENST00000393897.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007457.hg.1 | ENST00000409471.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007457.hg.1 | NM_001008387.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007457.hg.1 | NM_198448.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007458.hg.1 | TC02000487.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007458.hg.1 | ENST00000272324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007458.hg.1 | ENST00000409471.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007458.hg.1 | NM_001008387.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007459.hg.1 | TC02000487.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007459.hg.1 | ENST00000272324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007459.hg.1 | ENST00000393897.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007459.hg.1 | ENST00000409471.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007459.hg.1 | NM_001008387.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007459.hg.1 | NM_198448.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | ENST00000272324.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | ENST00000272324.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | ENST00000272324.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | ENST00000272324.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | ENST00000272324.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | ENST00000272324.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | ENST00000272324.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | ENST00000272324.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | ENST00000272324.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | ENST00000272324.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | ENST00000393897.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | ENST00000393897.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | ENST00000393897.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | ENST00000393897.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | ENST00000393897.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | ENST00000393897.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | ENST00000393897.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | ENST00000393897.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | ENST00000393897.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | ENST00000409471.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | ENST00000409471.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | ENST00000409471.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | ENST00000409471.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | ENST00000409471.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | ENST00000409471.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | ENST00000409471.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | ENST00000490944.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | ENST00000490944.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | ENST00000490944.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | ENST00000490944.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | ENST00000490944.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | ENST00000498312.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | ENST00000498312.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | ENST00000498312.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | ENST00000498312.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | ENST00000498312.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | ENST00000498312.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | NM_001008387.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | NM_001008387.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | NM_001008387.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | NM_001008387.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | NM_001008387.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | NM_001008387.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | NM_001008387.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | NM_001008387.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | NM_001008387.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | NM_001008387.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | NM_001270040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | NM_001270040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | NM_001270040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | NM_001270040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | NM_001270040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | NM_001270040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | NM_001270040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | NM_001270040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | NM_001270040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | NM_198448.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | NM_198448.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | NM_198448.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | NM_198448.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | NM_198448.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | NM_198448.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | NM_198448.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | NM_198448.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008167.hg.1 | NM_198448.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007453.hg.1 | TC02000487.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007453.hg.1 | ENST00000272324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007453.hg.1 | ENST00000393897.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007453.hg.1 | NM_001008387.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007453.hg.1 | NM_198448.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156585.hg.1 | ENST00000272324.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156585.hg.1 | ENST00000393897.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156585.hg.1 | NM_001008387.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156585.hg.1 | NM_001270040.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156585.hg.1 | NM_198448.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156585.hg.1 | ENST00000272324.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156585.hg.1 | ENST00000393897.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156585.hg.1 | NM_001008387.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156585.hg.1 | NM_001270040.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156585.hg.1 | NM_198448.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156585.hg.1 | TC0200008167.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | ENST00000272324.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | ENST00000272324.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | ENST00000272324.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | ENST00000272324.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | ENST00000272324.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | ENST00000272324.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | ENST00000272324.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | ENST00000272324.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | ENST00000272324.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | ENST00000272324.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | ENST00000393897.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | ENST00000393897.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | ENST00000393897.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | ENST00000393897.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | ENST00000393897.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | ENST00000393897.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | ENST00000393897.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | ENST00000393897.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | ENST00000393897.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | ENST00000409471.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | ENST00000409471.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | ENST00000409471.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | ENST00000409471.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | ENST00000409471.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | ENST00000409471.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | ENST00000409471.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | ENST00000490944.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | ENST00000490944.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | ENST00000490944.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | ENST00000490944.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | ENST00000490944.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | ENST00000498312.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | ENST00000498312.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | ENST00000498312.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | ENST00000498312.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | ENST00000498312.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | ENST00000498312.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | NM_001008387.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | NM_001008387.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | NM_001008387.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | NM_001008387.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | NM_001008387.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | NM_001008387.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | NM_001008387.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | NM_001008387.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | NM_001008387.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | NM_001008387.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | NM_001270040.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | NM_001270040.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | NM_001270040.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | NM_001270040.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | NM_001270040.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | NM_001270040.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | NM_001270040.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | NM_001270040.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | NM_001270040.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | NM_198448.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | NM_198448.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | NM_198448.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | NM_198448.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | NM_198448.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | NM_198448.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | NM_198448.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | NM_198448.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008167.hg.1 | NM_198448.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156587.hg.1 | ENST00000272324.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156587.hg.1 | ENST00000393897.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156587.hg.1 | ENST00000409471.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156587.hg.1 | ENST00000498312.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156587.hg.1 | NM_001008387.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156587.hg.1 | NM_001270040.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156587.hg.1 | NM_198448.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156587.hg.1 | ENST00000272324.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156587.hg.1 | ENST00000393897.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156587.hg.1 | ENST00000409471.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156587.hg.1 | ENST00000498312.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156587.hg.1 | NM_001008387.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156587.hg.1 | NM_001270040.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156587.hg.1 | NM_198448.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156587.hg.1 | TC0200008167.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200156589.hg.1 | ENST00000272324.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156589.hg.1 | ENST00000409471.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156589.hg.1 | ENST00000498312.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156589.hg.1 | NM_001008387.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156589.hg.1 | NM_001270040.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156589.hg.1 | ENST00000272324.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156589.hg.1 | ENST00000409471.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156589.hg.1 | ENST00000498312.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156589.hg.1 | NM_001008387.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156589.hg.1 | NM_001270040.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156589.hg.1 | TC0200008167.hg.1 | Clariom D TC |

REG3G
Genomic Map