Assay Details
| Gene Symbol: CAPN14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145122.1 | NP_001138594.1 | 10 - 11 | 1257 | 66 | |
| NM_001321270.1 | NP_001308199.1 | 10 - 11 | 1321 | 66 | ||
| XM_011532863.2 | XP_011531165.1 | 11 - 12 | 1418 | 66 | ||
| XM_011532864.2 | XP_011531166.1 | 11 - 12 | 1455 | 66 | ||
| XM_011532865.1 | XP_011531167.1 | 11 - 12 | 1359 | 66 | ||
| XM_011532866.2 | XP_011531168.1 | 9 - 10 | 1126 | 66 | ||
| GenBank mRNA | AK092257.1 | - | 10 - 11 | 1222 | 66 | |
| AK296910.1 | - | 12 - 13 | 1225 | 66 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001715.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02027686.hg.1 | TC02001715.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02027686.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02027686.hg.1 | ENST00000444918.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02027686.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02027682.hg.1 | TC02001715.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02027682.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02027682.hg.1 | ENST00000444918.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02027682.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200188828.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200188828.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200188828.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200188828.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200188828.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200188828.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200188828.hg.1 | TC0200012168.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200188830.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200188830.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200188830.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200188830.hg.1 | ENST00000398824.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200188830.hg.1 | ENST00000403897.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200188830.hg.1 | NM_001145122.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200188830.hg.1 | TC0200012168.hg.1 | Clariom D TC |

CAPN14
Genomic Map