Assay Details
Gene Symbol: GPATCH11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322249.1 | NP_001309178.1 | 3 - 4 | 416 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0300011548.hg.1 | ENST00000469247.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000473067.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000473067.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000473067.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000473067.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | ENST00000473067.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_174931.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_174931.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_174931.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_174931.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_174931.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_174931.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_174931.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_174931.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_174931.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_174931.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_174931.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007249.hg.1 | NM_174931.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000281932.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000409774.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000473067.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000473067.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000473067.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000473067.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | ENST00000473067.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_001278505.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_174931.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_174931.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_174931.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_174931.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_174931.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_174931.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_174931.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_174931.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_174931.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_174931.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_174931.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007249.hg.1 | NM_174931.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011548.hg.1 | ENST00000469247.1 | Ensembl |
