Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010573.hg.1 |
ENST00000558621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010573.hg.1 |
ENST00000558621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010573.hg.1 |
ENST00000558621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010573.hg.1 |
ENST00000558621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010573.hg.1 |
ENST00000558621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010573.hg.1 |
ENST00000558621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010573.hg.1 |
ENST00000558621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010573.hg.1 |
ENST00000558621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010573.hg.1 |
ENST00000559955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010573.hg.1 |
ENST00000559955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010573.hg.1 |
NR_120324.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010573.hg.1 |
NR_120324.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010573.hg.1 |
NR_120324.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010573.hg.1 |
NR_120324.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010573.hg.1 |
NR_120324.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500179489.hg.1 |
ENST00000558621.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500179489.hg.1 |
ENST00000559955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500179489.hg.1 |
NR_120324.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500179489.hg.1 |
ENST00000558621.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500179489.hg.1 |
ENST00000559955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500179489.hg.1 |
NR_120324.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500179489.hg.1 |
TC1500010573.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1500010573.hg.1 |
ENST00000558621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010573.hg.1 |
ENST00000558621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010573.hg.1 |
ENST00000558621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010573.hg.1 |
ENST00000558621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010573.hg.1 |
ENST00000558621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010573.hg.1 |
ENST00000558621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010573.hg.1 |
ENST00000558621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010573.hg.1 |
ENST00000558621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010573.hg.1 |
ENST00000559955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010573.hg.1 |
ENST00000559955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010573.hg.1 |
NR_120324.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010573.hg.1 |
NR_120324.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010573.hg.1 |
NR_120324.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010573.hg.1 |
NR_120324.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010573.hg.1 |
NR_120324.1 |
Refseq |