| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011442.hg.1 |
ENST00000375911.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011442.hg.1 |
ENST00000375911.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011442.hg.1 |
ENST00000375911.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011442.hg.1 |
ENST00000375911.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011442.hg.1 |
NM_021184.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011442.hg.1 |
NM_021184.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011442.hg.1 |
NM_021184.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011442.hg.1 |
NM_021184.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007603.hg.1 |
ENST00000422049.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007603.hg.1 |
ENST00000422049.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011442.hg.1 |
ENST00000375911.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011442.hg.1 |
ENST00000375911.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011442.hg.1 |
ENST00000375911.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011442.hg.1 |
ENST00000375911.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011442.hg.1 |
NM_021184.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011442.hg.1 |
NM_021184.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011442.hg.1 |
NM_021184.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011442.hg.1 |
NM_021184.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007603.hg.1 |
ENST00000422049.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007603.hg.1 |
ENST00000422049.1 |
Ensembl |