Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009436.hg.1 |
ENST00000524962.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009436.hg.1 |
ENST00000524962.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009436.hg.1 |
ENST00000524962.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009436.hg.1 |
ENST00000524962.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009436.hg.1 |
ENST00000529072.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009436.hg.1 |
ENST00000529072.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009436.hg.1 |
ENST00000529072.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009436.hg.1 |
ENST00000529072.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009436.hg.1 |
ENST00000529072.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009436.hg.1 |
ENST00000531193.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009436.hg.1 |
ENST00000531193.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009436.hg.1 |
ENST00000531193.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009436.hg.1 |
ENST00000531193.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009436.hg.1 |
ENST00000531193.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009436.hg.1 |
ENST00000531193.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009436.hg.1 |
ENST00000531193.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009436.hg.1 |
ENST00000531193.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009436.hg.1 |
ENST00000531193.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009436.hg.1 |
ENST00000531193.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009436.hg.1 |
ENST00000524962.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009436.hg.1 |
ENST00000524962.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009436.hg.1 |
ENST00000524962.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009436.hg.1 |
ENST00000524962.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009436.hg.1 |
ENST00000529072.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009436.hg.1 |
ENST00000529072.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009436.hg.1 |
ENST00000529072.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009436.hg.1 |
ENST00000529072.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009436.hg.1 |
ENST00000529072.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009436.hg.1 |
ENST00000531193.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009436.hg.1 |
ENST00000531193.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009436.hg.1 |
ENST00000531193.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009436.hg.1 |
ENST00000531193.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009436.hg.1 |
ENST00000531193.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009436.hg.1 |
ENST00000531193.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009436.hg.1 |
ENST00000531193.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009436.hg.1 |
ENST00000531193.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009436.hg.1 |
ENST00000531193.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009436.hg.1 |
ENST00000531193.1 |
Ensembl |