Assay Details
Gene Symbol: LOC338667 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017018652.1 | XP_016874141.1 | 3 - 4 | 1608 | 126 | |
GenBank mRNA | AB231780.1 | - | 3 - 4 | 682 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08098086 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08098057 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08098050 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08098051 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08098055 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08098080 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28525823 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08098061 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28525839 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08098083 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08098060 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08098064 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08098084 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08098082 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08098076 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08098078 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08098070 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16513461 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08098066 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08098074 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08098058 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16513459 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08098052 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08098071 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08098072 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28525829 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08098063 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28525815 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 481610 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 724993 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 125303 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 44831 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P19407071 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P11286027 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P19407071 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P11283620 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P340852 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P19407071 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT HG-U133+ PM Array Plate | 1564786_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC1100009436.hg.1 | ENST00000524962.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009436.hg.1 | ENST00000524962.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009436.hg.1 | ENST00000524962.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009436.hg.1 | ENST00000524962.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009436.hg.1 | ENST00000529072.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009436.hg.1 | ENST00000529072.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009436.hg.1 | ENST00000529072.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009436.hg.1 | ENST00000529072.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009436.hg.1 | ENST00000529072.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009436.hg.1 | ENST00000531193.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009436.hg.1 | ENST00000531193.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009436.hg.1 | ENST00000531193.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009436.hg.1 | ENST00000531193.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009436.hg.1 | ENST00000531193.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009436.hg.1 | ENST00000531193.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009436.hg.1 | ENST00000531193.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009436.hg.1 | ENST00000531193.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009436.hg.1 | ENST00000531193.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009436.hg.1 | ENST00000531193.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009436.hg.1 | ENST00000524962.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009436.hg.1 | ENST00000524962.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009436.hg.1 | ENST00000524962.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009436.hg.1 | ENST00000524962.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009436.hg.1 | ENST00000529072.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009436.hg.1 | ENST00000529072.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009436.hg.1 | ENST00000529072.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009436.hg.1 | ENST00000529072.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009436.hg.1 | ENST00000529072.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009436.hg.1 | ENST00000531193.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009436.hg.1 | ENST00000531193.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009436.hg.1 | ENST00000531193.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009436.hg.1 | ENST00000531193.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009436.hg.1 | ENST00000531193.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009436.hg.1 | ENST00000531193.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009436.hg.1 | ENST00000531193.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009436.hg.1 | ENST00000531193.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009436.hg.1 | ENST00000531193.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009436.hg.1 | ENST00000531193.1 | Ensembl |
Human Genome U133 Plus 2.0 Array | 1564786_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 1564786_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564786_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564786_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564786_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564786_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564786_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564786_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564786_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564786_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564786_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564786_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564786_at10 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1773558 | - | - |
HumanHT-12 v4 Expression BeadChip | ILMN_3251061 | - | - |
HumanHT-12 v4 Expression BeadChip | ILMN_3250204 | - | - |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1674185 | - | - |
Sentrix Human-6 Expression BeadChip | GI_37540736-S | BC043578.1 | human_transcriptome |
Illumina human-6 v1.0 expression beadchip | 0003440717 | - | - |
Illumina humanRef-8 v1.0 expression beadchip | 0003440717 | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1674185 | - | - |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1773558 | - | - |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1773558 | - | - |
