Assay Details
| Gene Symbol: ST3GAL4-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033839.1 | 1 - 2 | 145 | 67 | ||
| GenBank mRNA | BF343672.1 | - | 1 - 2 | 145 | 67 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11002434.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3708658 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 7132 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3708658 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P19408176 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P19408172 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02541672 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02541678 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 230999_PM_at | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 227925_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029158.hg.1 | ENST00000499602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029158.hg.1 | ENST00000501321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029158.hg.1 | NR_033839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029158.hg.1 | TC11002434.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012744.hg.1 | ENST00000626810.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012744.hg.1 | ENST00000626810.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012744.hg.1 | ENST00000626810.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012744.hg.1 | ENST00000626810.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012744.hg.1 | ENST00000629441.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012744.hg.1 | ENST00000629441.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012744.hg.1 | ENST00000629441.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012744.hg.1 | ENST00000629441.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012744.hg.1 | ENST00000629441.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012744.hg.1 | ENST00000629441.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012744.hg.1 | NR_033839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012744.hg.1 | NR_033839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012744.hg.1 | NR_033839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012744.hg.1 | NR_033839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029155.hg.1 | ENST00000501321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029155.hg.1 | NR_033839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029155.hg.1 | TC11002434.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029159.hg.1 | ENST00000499602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029159.hg.1 | NR_033839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029159.hg.1 | TC11002434.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1100197953.hg.1 | ENST00000629441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197953.hg.1 | NR_033839.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100197953.hg.1 | ENST00000629441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197953.hg.1 | NR_033839.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100197953.hg.1 | TC1100012744.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100012744.hg.1 | ENST00000626810.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012744.hg.1 | ENST00000626810.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012744.hg.1 | ENST00000626810.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012744.hg.1 | ENST00000626810.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012744.hg.1 | ENST00000629441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012744.hg.1 | ENST00000629441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012744.hg.1 | ENST00000629441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012744.hg.1 | ENST00000629441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012744.hg.1 | ENST00000629441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012744.hg.1 | ENST00000629441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012744.hg.1 | NR_033839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012744.hg.1 | NR_033839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012744.hg.1 | NR_033839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012744.hg.1 | NR_033839.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100197957.hg.1 | ENST00000626810.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197957.hg.1 | ENST00000629441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197957.hg.1 | NR_033839.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100197957.hg.1 | ENST00000626810.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197957.hg.1 | ENST00000629441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197957.hg.1 | NR_033839.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100197957.hg.1 | TC1100012744.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100197952.hg.1 | ENST00000629441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197952.hg.1 | NR_033839.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100197952.hg.1 | ENST00000629441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197952.hg.1 | NR_033839.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100197952.hg.1 | TC1100012744.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100197956.hg.1 | ENST00000626810.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197956.hg.1 | ENST00000629441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197956.hg.1 | NR_033839.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100197956.hg.1 | ENST00000626810.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197956.hg.1 | ENST00000629441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197956.hg.1 | NR_033839.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100197956.hg.1 | TC1100012744.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 1555974_PM_a_at | - | - |
| Human Genome U133 Plus 2.0 Array | 230999_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1555973_at | - | - |
| Human Genome U133 Plus 2.0 Array | 227925_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 230999_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1555973_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1555974_a_at | - | - |
| Human Genome U133 Plus 2.0 Array | 227925_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227925_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230999_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227925_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227925_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227925_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230999_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227925_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227925_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230999_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230999_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230999_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227925_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227925_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227925_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227925_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230999_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230999_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230999_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230999_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230999_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230999_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227925_at8 | - | - |
| Human Genome U95 Set | 43011_at | - | - |
| Human Genome U95 Set | 46960_PM_at | - | - |
| Human Genome U95 Set | 46960_at | - | - |
| Human Genome U95 Set | 43011_PM_at | - | - |
| Human Genome U133 Set | 227925_PM_at | - | - |
| Human Genome U133 Set | 230999_at | - | - |
| Human Genome U133 Set | 230999_PM_at | - | - |
| Human Genome U133 Set | 227925_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 233892 | - | - |

ST3GAL4-AS1
Genomic Map