Assay Details
| Gene Symbol: LINC00535 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033858.1 | 9 - 10 | 1420 | 58 | ||
| GenBank mRNA | BC037909.2 | - | 9 - 10 | 1420 | 58 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chromosome 8 open reading frame 87 | C8orf87 | -195.7kb | Sense | Chr. 8 - 93134095 - 93166850 on NCBI Build GRCh38 | |
| Downstream | family with sequence similarity 92 member A1 | FAM92A1 | 309.6kb | Anti-sense | Chr. 8 - 93700252 - 93732041 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08001425.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800011109.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011109.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011109.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011109.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011109.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011109.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011109.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011109.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011109.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011109.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011109.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011109.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011109.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011109.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011109.hg.1 | ENST00000517785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011109.hg.1 | ENST00000517785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011109.hg.1 | ENST00000520096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011109.hg.1 | ENST00000520096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011109.hg.1 | ENST00000520096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011109.hg.1 | ENST00000520096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011109.hg.1 | ENST00000520096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011109.hg.1 | ENST00000520096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011109.hg.1 | ENST00000520096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011109.hg.1 | ENST00000520096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011109.hg.1 | ENST00000520096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011109.hg.1 | ENST00000520096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011109.hg.1 | ENST00000520096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011109.hg.1 | ENST00000520096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011109.hg.1 | ENST00000520513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011109.hg.1 | ENST00000520513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011109.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011109.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011109.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011109.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011109.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011109.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011109.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011109.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011109.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011109.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011109.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011109.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011109.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011109.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08018499.hg.1 | TC08001425.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08018499.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08018499.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08018508.hg.1 | TC08001425.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08018508.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08018508.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08018498.hg.1 | TC08001425.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08018498.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08018498.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08018504.hg.1 | TC08001425.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08018504.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08018504.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08018503.hg.1 | TC08001425.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08018503.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08018503.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08018506.hg.1 | TC08001425.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08018506.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08018506.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08018500.hg.1 | TC08001425.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08018500.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08018500.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08018502.hg.1 | TC08001425.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08018502.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08018502.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08018505.hg.1 | TC08001425.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08018505.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08018505.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08018507.hg.1 | TC08001425.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08018507.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08018507.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800175682.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800175682.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800175682.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800175682.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800175682.hg.1 | TC0800011109.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800175689.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800175689.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800175689.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800175689.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800175689.hg.1 | TC0800011109.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800175693.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800175693.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800175693.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800175693.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800175693.hg.1 | TC0800011109.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800175687.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800175687.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800175687.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800175687.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800175687.hg.1 | TC0800011109.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800175690.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800175690.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800175690.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800175690.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800175690.hg.1 | TC0800011109.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800175691.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800175691.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800175691.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800175691.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800175691.hg.1 | TC0800011109.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800175683.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800175683.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800175683.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800175683.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800175683.hg.1 | TC0800011109.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800175692.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800175692.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800175692.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800175692.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800175692.hg.1 | TC0800011109.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800175681.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800175681.hg.1 | ENST00000520096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800175681.hg.1 | ENST00000520096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800175681.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800175681.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800175681.hg.1 | ENST00000520096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800175681.hg.1 | ENST00000520096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800175681.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800175681.hg.1 | TC0800011109.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800011109.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011109.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011109.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011109.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011109.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011109.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011109.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011109.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011109.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011109.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011109.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011109.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011109.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011109.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011109.hg.1 | ENST00000517785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011109.hg.1 | ENST00000517785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011109.hg.1 | ENST00000520096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011109.hg.1 | ENST00000520096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011109.hg.1 | ENST00000520096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011109.hg.1 | ENST00000520096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011109.hg.1 | ENST00000520096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011109.hg.1 | ENST00000520096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011109.hg.1 | ENST00000520096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011109.hg.1 | ENST00000520096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011109.hg.1 | ENST00000520096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011109.hg.1 | ENST00000520096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011109.hg.1 | ENST00000520096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011109.hg.1 | ENST00000520096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011109.hg.1 | ENST00000520513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011109.hg.1 | ENST00000520513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011109.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011109.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011109.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011109.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011109.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011109.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011109.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011109.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011109.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011109.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011109.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011109.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011109.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011109.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800175688.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800175688.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800175688.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800175688.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800175688.hg.1 | TC0800011109.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800175685.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800175685.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800175685.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800175685.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800175685.hg.1 | TC0800011109.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800175686.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800175686.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800175686.hg.1 | ENST00000501400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800175686.hg.1 | NR_033858.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800175686.hg.1 | TC0800011109.hg.1 | Clariom D TC |

LINC00535
Genomic Map