Assay Details
| Gene Symbol: LOC101929473 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120578.1 | 2 - 3 | 290 | 101 | ||
| GenBank mRNA | AK095187.1 | - | 2 - 3 | 290 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929473| Gene Symbol: LOC101929473 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120578.1 | 2 - 3 | 290 | 101 | ||
| GenBank mRNA | AK095187.1 | - | 2 - 3 | 290 | 101 | |
Set Membership: |
|