Assay Details
Gene Symbol: LINC00996 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_034033.1 | 2 - 3 | 790 | 82 | ||
GenBank mRNA | AK097158.1 | - | 2 - 3 | 692 | 82 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC07001001.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0700009674.hg.1 | ENST00000477392.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009674.hg.1 | ENST00000477392.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009674.hg.1 | ENST00000477392.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009674.hg.1 | ENST00000477392.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009674.hg.1 | ENST00000477392.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009674.hg.1 | ENST00000477392.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009674.hg.1 | ENST00000477392.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009674.hg.1 | ENST00000486954.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009674.hg.1 | ENST00000486954.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009674.hg.1 | ENST00000486954.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009674.hg.1 | NR_034033.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009674.hg.1 | NR_034033.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009674.hg.1 | NR_034033.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009674.hg.1 | NR_034033.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009674.hg.1 | NR_034033.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009674.hg.1 | NR_034033.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009674.hg.1 | NR_034033.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07015241.hg.1 | TC07001001.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07015241.hg.1 | ENST00000477392.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07015241.hg.1 | NR_034033.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07015238.hg.1 | TC07001001.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07015238.hg.1 | ENST00000477392.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07015238.hg.1 | NR_034033.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07015242.hg.1 | TC07001001.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07015242.hg.1 | ENST00000477392.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07015242.hg.1 | NR_034033.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07015243.hg.1 | TC07001001.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07015243.hg.1 | ENST00000477392.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07015243.hg.1 | NR_034033.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07015239.hg.1 | TC07001001.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07015239.hg.1 | ENST00000477392.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07015239.hg.1 | NR_034033.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009674.hg.1 | ENST00000477392.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009674.hg.1 | ENST00000477392.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009674.hg.1 | ENST00000477392.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009674.hg.1 | ENST00000477392.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009674.hg.1 | ENST00000477392.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009674.hg.1 | ENST00000477392.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009674.hg.1 | ENST00000477392.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009674.hg.1 | ENST00000486954.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009674.hg.1 | ENST00000486954.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009674.hg.1 | ENST00000486954.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009674.hg.1 | NR_034033.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009674.hg.1 | NR_034033.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009674.hg.1 | NR_034033.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009674.hg.1 | NR_034033.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009674.hg.1 | NR_034033.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009674.hg.1 | NR_034033.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009674.hg.1 | NR_034033.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700168701.hg.1 | ENST00000477392.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700168701.hg.1 | NR_034033.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700168701.hg.1 | ENST00000477392.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700168701.hg.1 | NR_034033.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700168701.hg.1 | TC0700009674.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700168694.hg.1 | ENST00000477392.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700168694.hg.1 | NR_034033.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700168694.hg.1 | ENST00000477392.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700168694.hg.1 | NR_034033.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700168694.hg.1 | TC0700009674.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700168696.hg.1 | ENST00000477392.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700168696.hg.1 | ENST00000486954.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700168696.hg.1 | NR_034033.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700168696.hg.1 | ENST00000477392.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700168696.hg.1 | ENST00000486954.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700168696.hg.1 | NR_034033.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700168696.hg.1 | TC0700009674.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700168704.hg.1 | ENST00000477392.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700168704.hg.1 | NR_034033.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700168704.hg.1 | ENST00000477392.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700168704.hg.1 | NR_034033.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700168704.hg.1 | TC0700009674.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700168700.hg.1 | ENST00000477392.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700168700.hg.1 | ENST00000486954.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700168700.hg.1 | NR_034033.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700168700.hg.1 | ENST00000477392.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700168700.hg.1 | ENST00000486954.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700168700.hg.1 | NR_034033.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700168700.hg.1 | TC0700009674.hg.1 | Clariom D TC |
