Assay Details
| Gene Symbol: LOC257396 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_034107.2 | 2 - 3 | 1099 | 104 | ||
| GenBank mRNA | AK097893.1 | - | 2 - 3 | 1084 | 104 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000219.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24100789 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24100798 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24100800 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24100805 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24100788 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24100796 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24100785 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24100794 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03928279 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03928271 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24100790 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24100795 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03928273 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24100803 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24100808 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24100797 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24100792 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24100802 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24100793 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15690537 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03928256 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24100791 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 227501 | AF141310.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 227501 | AK124646.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 227501 | BC033656.2 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 227501 | BC065031.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 751482 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 401770 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3231267 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3231262 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_32_P92563 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 25066 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 21126 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 33846 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_18_P15188521 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3231262 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3231262 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3231267 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_32_P92563 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P17117463 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_32_P92563 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P17117463 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P15188521 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P17117467 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P201641 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P92563 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P17117463 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 236076_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007370.hg.1 | ENST00000499459.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007370.hg.1 | ENST00000499459.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007370.hg.1 | ENST00000499459.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007370.hg.1 | ENST00000499459.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007370.hg.1 | ENST00000499459.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007370.hg.1 | ENST00000499459.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007370.hg.1 | ENST00000499459.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007370.hg.1 | ENST00000499459.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007370.hg.1 | ENST00000499459.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007370.hg.1 | ENST00000502171.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007370.hg.1 | ENST00000502171.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007370.hg.1 | ENST00000502171.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007370.hg.1 | ENST00000502171.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007370.hg.1 | ENST00000502171.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007370.hg.1 | ENST00000502171.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007370.hg.1 | ENST00000502171.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007370.hg.1 | ENST00000512301.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007370.hg.1 | ENST00000512301.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007370.hg.1 | ENST00000512301.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007370.hg.1 | NR_034107.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007370.hg.1 | NR_034107.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007370.hg.1 | NR_034107.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007370.hg.1 | NR_034107.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007370.hg.1 | NR_034107.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007370.hg.1 | NR_034107.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007370.hg.1 | NR_034107.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007370.hg.1 | NR_104654.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007370.hg.1 | NR_104654.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007370.hg.1 | NR_104654.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007370.hg.1 | NR_104654.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007370.hg.1 | NR_104654.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002718.hg.1 | TC05000219.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002718.hg.1 | ENST00000502171.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002718.hg.1 | NR_034107.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002719.hg.1 | TC05000219.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002719.hg.1 | ENST00000502171.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002719.hg.1 | NR_034107.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500149835.hg.1 | ENST00000499459.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500149835.hg.1 | ENST00000502171.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500149835.hg.1 | ENST00000512301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500149835.hg.1 | NR_034107.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500149835.hg.1 | NR_104654.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500149835.hg.1 | ENST00000499459.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500149835.hg.1 | ENST00000502171.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500149835.hg.1 | ENST00000512301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500149835.hg.1 | NR_034107.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500149835.hg.1 | NR_104654.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500149835.hg.1 | TC0500007370.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500149836.hg.1 | ENST00000499459.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500149836.hg.1 | ENST00000502171.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500149836.hg.1 | NR_034107.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500149836.hg.1 | NR_104654.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500149836.hg.1 | ENST00000499459.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500149836.hg.1 | ENST00000502171.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500149836.hg.1 | NR_034107.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500149836.hg.1 | NR_104654.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500149836.hg.1 | TC0500007370.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500007370.hg.1 | ENST00000499459.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007370.hg.1 | ENST00000499459.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007370.hg.1 | ENST00000499459.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007370.hg.1 | ENST00000499459.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007370.hg.1 | ENST00000499459.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007370.hg.1 | ENST00000499459.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007370.hg.1 | ENST00000499459.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007370.hg.1 | ENST00000499459.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007370.hg.1 | ENST00000499459.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007370.hg.1 | ENST00000502171.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007370.hg.1 | ENST00000502171.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007370.hg.1 | ENST00000502171.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007370.hg.1 | ENST00000502171.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007370.hg.1 | ENST00000502171.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007370.hg.1 | ENST00000502171.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007370.hg.1 | ENST00000502171.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007370.hg.1 | ENST00000512301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007370.hg.1 | ENST00000512301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007370.hg.1 | ENST00000512301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007370.hg.1 | NR_034107.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007370.hg.1 | NR_034107.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007370.hg.1 | NR_034107.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007370.hg.1 | NR_034107.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007370.hg.1 | NR_034107.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007370.hg.1 | NR_034107.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007370.hg.1 | NR_034107.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007370.hg.1 | NR_104654.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007370.hg.1 | NR_104654.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007370.hg.1 | NR_104654.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007370.hg.1 | NR_104654.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007370.hg.1 | NR_104654.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500149837.hg.1 | ENST00000499459.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500149837.hg.1 | ENST00000502171.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500149837.hg.1 | NR_034107.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500149837.hg.1 | NR_104654.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500149837.hg.1 | ENST00000499459.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500149837.hg.1 | ENST00000502171.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500149837.hg.1 | NR_034107.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500149837.hg.1 | NR_104654.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500149837.hg.1 | TC0500007370.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 236076_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 236076_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 236076_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 236076_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 236076_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 236076_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 236076_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 236076_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 236076_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 236076_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 236076_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 236076_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 236076_at8 | - | - |
| Human Genome U95 Set | 65047_at | - | - |
| Human Genome U95 Set | 65047_PM_at | - | - |
| Human Genome U133 Set | 236076_at | - | - |
| Human Genome U133 Set | 236076_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A055205 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A055205 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 165703 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3215374 | BC041894.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3215374 | NR_034107.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3215374 | AK097893.1 | human_transcriptome |

LOC257396
Genomic Map