Assay Details
| Gene Symbol: LINC00928 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK058110.1 | - | 2 - 3 | 587 | 135 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Rh family C glycoprotein | RHCG |
C15orf6, PDRC2, RHGK, SLC42A3 |
-18.0kb | Sense | Chr. 15 - 89471407 - 89496613 on NCBI Build GRCh38 |
| Downstream | TOPBP1 interacting checkpoint and replication regulator | TICRR |
C15orf42, SLD3, Treslin |
58.0kb | Anti-sense | Chr. 15 - 89575587 - 89628023 on NCBI Build GRCh38 |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | ENST00000559227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | ENST00000559227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | ENST00000559227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | ENST00000559227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | ENST00000559227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | ENST00000559967.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | ENST00000559967.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | ENST00000559967.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | ENST00000559967.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | ENST00000561108.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | ENST00000561108.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | ENST00000561108.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | ENST00000561108.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | ENST00000561108.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | ENST00000561108.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | ENST00000561108.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | ENST00000561108.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | ENST00000561108.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | ENST00000561108.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | ENST00000561201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | ENST00000561201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | ENST00000561201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | ENST00000561201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | ENST00000561201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | ENST00000561201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | ENST00000561201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | ENST00000561201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | ENST00000561201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | ENST00000561201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | ENST00000561201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | ENST00000561201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | ENST00000561201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | ENST00000561201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | NR_027074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | NR_027074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | NR_027074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | NR_027074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | NR_027074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | NR_027074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | NR_027074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | NR_027074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | NR_027075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | NR_027075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | NR_027075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | NR_027075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | NR_027075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | NR_027075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | NR_027075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | NR_027075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | NR_027076.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | NR_027076.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | NR_027076.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | NR_027076.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | NR_027076.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | NR_027077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | NR_027077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | NR_027077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | NR_027077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | NR_027077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010392.hg.1 | NR_027077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | ENST00000559227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | ENST00000559227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | ENST00000559227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | ENST00000559227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | ENST00000559227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | ENST00000559967.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | ENST00000559967.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | ENST00000559967.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | ENST00000559967.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | ENST00000561108.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | ENST00000561108.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | ENST00000561108.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | ENST00000561108.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | ENST00000561108.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | ENST00000561108.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | ENST00000561108.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | ENST00000561108.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | ENST00000561108.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | ENST00000561108.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | ENST00000561201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | ENST00000561201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | ENST00000561201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | ENST00000561201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | ENST00000561201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | ENST00000561201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | ENST00000561201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | ENST00000561201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | ENST00000561201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | ENST00000561201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | ENST00000561201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | ENST00000561201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | ENST00000561201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | ENST00000561201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | NR_027074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | NR_027074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | NR_027074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | NR_027074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | NR_027074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | NR_027074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | NR_027074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | NR_027074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | NR_027075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | NR_027075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | NR_027075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | NR_027075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | NR_027075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | NR_027075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | NR_027075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | NR_027075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | NR_027076.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | NR_027076.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | NR_027076.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | NR_027076.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | NR_027076.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | NR_027077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | NR_027077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | NR_027077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | NR_027077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | NR_027077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010392.hg.1 | NR_027077.1 | Refseq |

LINC00928
Genomic Map