Assay Details
| Gene Symbol: PLEKHG4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_052909.3 | NP_443141.3 | 15 - 16 | 3208 | 65 | |
| XM_006714446.3 | XP_006714509.1 | 17 - 18 | 4346 | 65 | ||
| XM_011513968.2 | XP_011512270.1 | 17 - 18 | 4343 | 65 | ||
| XM_011513969.2 | XP_011512271.1 | 17 - 18 | 4223 | 65 | ||
| XM_011513970.1 | XP_011512272.1 | 17 - 18 | 3935 | 65 | ||
| XM_011513971.2 | XP_011512273.1 | 17 - 18 | 4291 | 65 | ||
| XM_017009112.1 | XP_016864601.1 | 17 - 18 | 4347 | 65 | ||
| GenBank mRNA | AB067496.2 | - | 15 - 16 | 3208 | 65 | |
| AK125163.1 | - | 4 - 5 | 1037 | 65 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000002.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05000022.hg.1 | TC05000002.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05000022.hg.1 | ENST00000283426.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05000022.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05000023.hg.1 | TC05000002.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05000023.hg.1 | ENST00000283426.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05000023.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500144081.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500144081.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500144081.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500144081.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500144081.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500144081.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500144081.hg.1 | TC0500006435.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500144082.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500144082.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500144082.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500144082.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500144082.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500144082.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500144082.hg.1 | TC0500006435.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |

PLEKHG4B
Genomic Map