Assay Details
| Gene Symbol: PLEKHG4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017009112.1 | XP_016864601.1 | 18 - 19 | 4529 | 67 | |
| GenBank mRNA | AK125163.1 | - | 5 - 6 | 1219 | 67 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |

PLEKHG4B
Genomic Map