Assay Details
| Gene Symbol: PLEKHG4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK125163.1 | - | 6 - 7 | 1410 | 87 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |

PLEKHG4B
Genomic Map