Assay Details
| Gene Symbol: LINC01250 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110228.1 | 1 - 2 | 194 | 73 | ||
| GenBank mRNA | AK095310.1 | - | 1 - 2 | 194 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01250| Gene Symbol: LINC01250 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110228.1 | 1 - 2 | 194 | 73 | ||
| GenBank mRNA | AK095310.1 | - | 1 - 2 | 194 | 73 | |
Set Membership: |
|