Assay Details
| Gene Symbol: LOC101927948 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110287.1 | 4 - 5 | 564 | 96 | ||
| GenBank mRNA | BC042517.1 | - | 4 - 5 | 564 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927948| Gene Symbol: LOC101927948 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110287.1 | 4 - 5 | 564 | 96 | ||
| GenBank mRNA | BC042517.1 | - | 4 - 5 | 564 | 96 | |
Set Membership: |
|