Assay Details
| Gene Symbol: LOC101927948 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110287.1 | 5 - 6 | 795 | 151 | ||
| GenBank mRNA | BC042517.1 | - | 5 - 6 | 795 | 151 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927948| Gene Symbol: LOC101927948 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110287.1 | 5 - 6 | 795 | 151 | ||
| GenBank mRNA | BC042517.1 | - | 5 - 6 | 795 | 151 | |
Set Membership: |
|