Assay Details
| Gene Symbol: SH2D6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201594.2 | NP_963888.2 | 11 - 12 | 1135 | 76 | |
| XM_017003842.1 | XP_016859331.1 | 6 - 7 | 4212 | 76 | ||
| XM_017003846.1 | XP_016859335.1 | 6 - 7 | 4158 | 76 | ||
| XM_017003858.1 | XP_016859347.1 | 11 - 12 | 1135 | 76 | ||
| XM_017003859.1 | XP_016859348.1 | 2 - 3 | 204 | 76 | ||
| GenBank mRNA | AK093312.1 | - | 11 - 12 | 1135 | 76 | |
| AY517502.1 | - | 11 - 12 | 1135 | 76 | ||
| BC110073.1 | - | 2 - 3 | 170 | 76 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AK093312.1,AY517502.1,BC110073.1,NM_201594.2,XM_017003858.1,XM_017003859.1 transcripts.
This assay is in the 5'UTR region of the AK093312.1,AY517502.1,BC110073.1,NM_201594.2,XM_017003858.1,XM_017003859.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000340326.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000340326.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000340326.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000340326.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000340326.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000340326.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000340326.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000340326.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000340326.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000469800.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000469800.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000469800.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000469800.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000469800.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000474753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000474753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000477170.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000477170.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000477170.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000477170.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000477170.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000477170.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000477170.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000477170.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000477170.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000481395.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000481395.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000481395.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000481395.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000481395.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000481395.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000481395.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000481395.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000481395.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000481395.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000481395.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000481395.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000481395.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000481395.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000481426.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000481426.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000481426.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000481426.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000481426.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000481426.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000481426.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000481426.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000481426.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000481426.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000481426.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000481426.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000481426.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000481426.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000488219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000488219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000488219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000488219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000488657.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000488657.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000488657.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000488657.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000488657.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000488657.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000488657.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000488657.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008253.hg.1 | ENST00000488657.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000340326.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000340326.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000340326.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000340326.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000340326.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000340326.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000340326.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000340326.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000340326.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000389938.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000469800.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000469800.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000469800.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000469800.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000469800.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000474753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000474753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000477170.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000477170.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000477170.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000477170.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000477170.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000477170.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000477170.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000477170.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000477170.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000481395.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000481395.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000481395.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000481395.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000481395.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000481395.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000481395.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000481395.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000481395.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000481395.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000481395.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000481395.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000481395.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000481395.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000481426.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000481426.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000481426.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000481426.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000481426.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000481426.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000481426.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000481426.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000481426.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000481426.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000481426.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000481426.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000481426.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000481426.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000488219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000488219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000488219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000488219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000488657.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000488657.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000488657.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000488657.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000488657.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000488657.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000488657.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000488657.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008253.hg.1 | ENST00000488657.3 | Ensembl |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 128309 | - | - |

SH2D6
Genomic Map