Assay Details
| Gene Symbol: DPY19L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015283.1 | NP_056098.1 | 14 - 15 | 1347 | 84 | |
| XM_011515245.2 | XP_011513547.1 | 14 - 15 | 1496 | 84 | ||
| XM_011515246.2 | XP_011513548.1 | 13 - 14 | 1409 | 84 | ||
| GenBank mRNA | AB020684.1 | - | 11 - 12 | 918 | 84 | |
| DQ287932.1 | - | 14 - 15 | 1347 | 84 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07001274.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700007129.hg.1 | ENST00000426647.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007129.hg.1 | ENST00000426647.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007129.hg.1 | ENST00000426647.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007129.hg.1 | ENST00000426647.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007129.hg.1 | ENST00000426647.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007129.hg.1 | ENST00000426647.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007129.hg.1 | ENST00000426647.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007129.hg.1 | ENST00000426647.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007129.hg.1 | ENST00000426647.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007129.hg.1 | NR_132360.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007129.hg.1 | NR_132360.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007129.hg.1 | NR_132360.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007129.hg.1 | NR_132360.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007129.hg.1 | NR_132360.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007129.hg.1 | NR_132360.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007129.hg.1 | NR_132360.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007129.hg.1 | NR_132360.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000428054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000428054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000446375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000446375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000446375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000446375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000446375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000446375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000446375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000446375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000462134.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000462134.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000462134.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000462134.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000462134.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000463538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000463538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000463538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000481923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000481923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000481923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000481923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000481923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000481923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000493989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000493989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000493989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000612226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000612226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000612226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000612226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007129.hg.1 | ENST00000426647.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007129.hg.1 | ENST00000426647.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007129.hg.1 | ENST00000426647.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007129.hg.1 | ENST00000426647.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007129.hg.1 | ENST00000426647.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007129.hg.1 | ENST00000426647.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007129.hg.1 | ENST00000426647.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007129.hg.1 | ENST00000426647.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007129.hg.1 | ENST00000426647.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007129.hg.1 | NR_132360.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007129.hg.1 | NR_132360.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007129.hg.1 | NR_132360.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007129.hg.1 | NR_132360.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007129.hg.1 | NR_132360.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007129.hg.1 | NR_132360.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007129.hg.1 | NR_132360.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007129.hg.1 | NR_132360.1 | Refseq |

DPY19L1
Genomic Map