Assay Details
| Gene Symbol: C1orf189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001010979.2 | NP_001010979.1 | 3 - 4 | 266 | 97 | |
| GenBank mRNA | BC127710.1 | - | 3 - 4 | 265 | 40127224 | 97 |
| BC130509.1 | - | 3 - 4 | 324 | 40146540 | 97 | |
| HY025946.1 | - | 3 - 4 | 266 | 97 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003289.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20576591 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20576581 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00519080 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20576590 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00519074 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00519070 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00519082 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00519071 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20576573 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00519077 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00519081 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20576586 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00519075 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20576593 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00519078 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20576594 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00519076 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20576582 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20576583 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 395256 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 209574 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3250944 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0878 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0877 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 54939 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P00167319 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P30314978 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_18_P10330769 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P30314950 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3250944 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3250944 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15130745 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20576581 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00519078 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00519070 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20576582 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00519081 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00519077 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20576573 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00519071 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20576586 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20576593 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00519074 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20576590 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00519072 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00519082 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20576594 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20576594 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00519082 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20576586 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20576573 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20576582 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15130745 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20576593 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00519074 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00519078 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00519077 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00519081 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00519071 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00519072 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20576590 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20576581 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00519070 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00519082 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00519079 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00519081 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00519070 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00519085 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00519084 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00519080 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00167317 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00167315 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P17846456 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P30314950 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00519075 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00519073 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00519124 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15130746 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00519002 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15130745 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00519079 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00519058 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00519056 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00519049 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00518998 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00519006 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00519011 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00519013 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00519047 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00519026 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00519025 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00519044 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00519034 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00519123 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15130741 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00519008 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00519116 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00519053 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00519004 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00519117 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00519051 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00519080 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00519022 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00519039 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00519036 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00519028 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00519018 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00519120 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00519015 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00519000 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00519084 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00519121 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00519042 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00519060 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00519082 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00519029 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00519032 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00519085 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00519077 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00519062 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00519030 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00519070 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P00167317 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P35313330 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_001010979_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7920491 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7920489 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7920488 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7920490 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7920487 | - | - |
| Affymetrix Human Genome U219 Array | 11737207_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11737207_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050727.hg.1 | TC01003289.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050727.hg.1 | ENST00000368525.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050727.hg.1 | NM_001010979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015901.hg.1 | ENST00000368525.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015901.hg.1 | ENST00000368525.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015901.hg.1 | ENST00000368525.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015901.hg.1 | ENST00000368525.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015901.hg.1 | ENST00000368525.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015901.hg.1 | ENST00000368525.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015901.hg.1 | NM_001010979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015901.hg.1 | NM_001010979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015901.hg.1 | NM_001010979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015901.hg.1 | NM_001010979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015901.hg.1 | NM_001010979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015901.hg.1 | NM_001010979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015901.hg.1 | NM_001010979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015901.hg.1 | NM_001010979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015901.hg.1 | NM_001010979.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100219728.hg.1 | ENST00000368525.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219728.hg.1 | NM_001010979.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100219728.hg.1 | ENST00000368525.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219728.hg.1 | NM_001010979.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100219728.hg.1 | TC0100015901.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100015901.hg.1 | ENST00000368525.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015901.hg.1 | ENST00000368525.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015901.hg.1 | ENST00000368525.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015901.hg.1 | ENST00000368525.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015901.hg.1 | ENST00000368525.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015901.hg.1 | ENST00000368525.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015901.hg.1 | NM_001010979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015901.hg.1 | NM_001010979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015901.hg.1 | NM_001010979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015901.hg.1 | NM_001010979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015901.hg.1 | NM_001010979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015901.hg.1 | NM_001010979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015901.hg.1 | NM_001010979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015901.hg.1 | NM_001010979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015901.hg.1 | NM_001010979.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100219725.hg.1 | NM_001010979.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100219725.hg.1 | NM_001010979.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100219725.hg.1 | TC0100015901.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100219727.hg.1 | ENST00000368525.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219727.hg.1 | NM_001010979.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100219727.hg.1 | ENST00000368525.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219727.hg.1 | NM_001010979.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100219727.hg.1 | TC0100015901.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100219724.hg.1 | NM_001010979.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100219724.hg.1 | NM_001010979.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100219724.hg.1 | TC0100015901.hg.1 | Clariom D TC |
| Human Genome U95 Set | 64986_at | - | - |
| Human Genome U95 Set | 64986_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A052662 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A052662 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 714113 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1790713 | NM_001010979.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1790713 | BC127710.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1790713 | BC130509.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1790713 | NM_001010979.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1790713 | BC127710.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1790713 | BC130509.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1790713 | NM_001010979.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1790713 | BC127710.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1790713 | BC130509.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1790713 | NM_001010979.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1790713 | BC127710.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1790713 | BC130509.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1790713 | NM_001010979.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1790713 | BC127710.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1790713 | BC130509.1 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_10361-S | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1790713 | NM_001010979.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1790713 | BC127710.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1790713 | BC130509.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1790713 | NM_001010979.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1790713 | BC127710.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1790713 | BC130509.1 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 6180228 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_10361 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1790713 | NM_001010979.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1790713 | BC127710.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1790713 | BC130509.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1790713 | NM_001010979.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1790713 | BC127710.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1790713 | BC130509.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1790713 | NM_001010979.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1790713 | BC127710.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1790713 | BC130509.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1790713 | NM_001010979.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1790713 | BC127710.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1790713 | BC130509.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1790713 | NM_001010979.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1790713 | BC127710.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1790713 | BC130509.1 | human_transcriptome |

C1orf189
Genomic Map